Skip to Content
Merck
All Photos(1)

Documents

A6719

Sigma-Aldrich

ω-Agatoxin IVA

≥90% (HPLC)

Synonym(s):

SNX 290

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C217H360N68O60S10
CAS Number:
Molecular Weight:
5202.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥90% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

Gene Information

Amino Acid Sequence

Lys-Lys-Lys-Cys-Ile-Ala-Lys-Asp-Tyr-Gly-Arg-Cys-Lys-Trp-Gly-Gly-Thr-Pro-Cys-Cys-Arg-Gly-Arg-Gly-Cys-Ile-Cys-Ser-Ile-Met-Gly-Thr-Asn-Cys-Glu-Cys-Lys-Pro-Arg-Leu-Ile-Met-Glu-Gly-Leu-Gly-Leu-Ala [Disulfide bridge: 4-20, 12-25, 19-36, 27-34]

General description

CACNA1A (calcium voltage-gated channel subunit alpha1 A) gene codes for the α1 subunit of neuronal CaV2.1 (P/Q-type) voltage-gated calcium channels. It is expressed throughout the central nervous system (CNS). CACNA1A gene is located on human chromosome 19p13.

Application

ω-Agatoxin IVA has been used in the manipulation of calcium influx. It has also been used in the study to characterize the spontaneous and evoked activity from murine ventral horn cultures on microelectrode arrays.

Biochem/physiol Actions

Mutations in the CACNA1A (calcium voltage-gated channel subunit alpha1 A) gene is responsible for episodic ataxia 2, familial hemiplegic migraine type 1 (FHM1) and spinocerebellar ataxia type 6 (SCA6).
Toxin found in the venom of the funnel web spider, Agelenopsis aptera. Potent, selective blocker of mammalian P-type voltage-dependent calcium channels.

Features and Benefits

This compound was developed by Pfizer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I M Mintz et al.
Nature, 355(6363), 827-829 (1992-02-27)
Voltage-dependent calcium channels mediate calcium entry into neurons, which is crucial for many processes in the brain including synaptic transmission, dendritic spiking, gene expression and cell death. Many types of calcium channels exist in mammalian brains, but high-affinity blockers are
Involvement of the CACNA1A gene containing region on 19p13 in migraine with and without aura
Terwindt G M, et al.
Neurology, 56(8), 1028-1032 (2001)
I M Mintz et al.
Neuron, 9(1), 85-95 (1992-07-01)
The peptide toxin omega-Aga-IVA blocked P-type Ca2+ channel current in rat Purkinje neurons (KD approximately 2 nM) but had no effect on identified T-type, L-type, or N-type currents in a variety of central and peripheral neurons. omega-Aga-IVA blocked a substantial
Charge-balanced biphasic electrical stimulation inhibits neurite extension of spiral ganglion neurons
Shen N, et al.
Neuroscience Letters, 624, 92-99 (2016)
New CACNA1A deletions are associated to migraine phenotypes
Grieco GS, et al.
The Journal of Headache and Pain, 19(1), 75-75 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service