Skip to Content
Merck

A6402

[Asn1, Val5]-Angiotensin II

≥97% (HPLC), angiotensin agonist, powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C49H70N14O11
CAS Number:
Molecular Weight:
1031.17
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

[Asn1, Val5]-Angiotensin II, ≥97% (HPLC)

Quality Level

assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CC(N)=O)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O

InChI

1S/C49H70N14O11/c1-26(2)39(61-42(67)33(12-8-18-55-49(52)53)57-41(66)32(50)23-38(51)65)45(70)58-34(20-29-14-16-31(64)17-15-29)43(68)62-40(27(3)4)46(71)59-35(22-30-24-54-25-56-30)47(72)63-19-9-13-37(63)44(69)60-36(48(73)74)21-28-10-6-5-7-11-28/h5-7,10-11,14-17,24-27,32-37,39-40,64H,8-9,12-13,18-23,50H2,1-4H3,(H2,51,65)(H,54,56)(H,57,66)(H,58,70)(H,59,71)(H,60,69)(H,61,67)(H,62,68)(H,73,74)(H4,52,53,55)/t32-,33-,34-,35-,36-,37-,39-,40-/m0/s1

InChI key

JYPVVOOBQVVUQV-CGHBYZBKSA-N

Gene Information

human ... AGTR1(185)
mouse ... AGTR1(11607)
rat ... AGTR1(24180)

General description

Angiotensin II is a renin-angiotensin system (RAS) effector molecule, which binds to angiotensin II type-1 (AT1R) and angiotensin II type-2 (AT2R) receptors. The gene is located on human chromosome 3q24. Angiotensin II is mainly expressed in vessel walls, lung and kidney.

Application

[Asn1, Val5]-Angiotensin II has been used:
  • to study the effects of ACTH, serotonin, Ang II, carbachol and elevated [K+] on catecholamine and serotonin release
  • to induce cyclophilin D (CypD)–dependent mitochondrial vascular superoxide (O2. - ) production in hypertension
  • to increase the homocysteine-induced ER stress-inducible, ubiquitin-like domain member 1 (HERPUD1) protein level in HeLa cells

Biochem/physiol Actions

Synthetic angiotensin agonist that is involved in the regulation of blood pressure, water consumption, and hydromineral balance in fish.
Angiotensin II functions as a vasoconstrictor and controls cardiovascular function. It maintains water-sodium homeostasis, modulates cellular proliferation and vascular remodelling of renin-angiotensin system. Angiotensin II has inflammatory effects.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Targeting Angiotensin II Type-1 Receptor (AT1R) Inhibits the Harmful Phenotype of Plasmodium-Specific CD8+ T Cells during Blood-Stage Malaria
Silva-Filho JL, et al.
Frontiers in Cellular and Infection Microbiology, 7, 42-42 (2017)
Control of catecholamine and serotonin release from the chromaffin tissue of the Atlantic hagfish
Bernier N and Perry S
The Journal of Experimental Biology, 199(11), 2485-2497 (1996)
The glu298asp polymorphism in the nitric oxide synthase 3 gene is associated with the risk of ischemic stroke in two large independent case-control studies
Berger K, et al.
Human Genetics, 121(2), 169-178 (2007)



Global Trade Item Number

SKUGTIN
A6402-1MG04061826149317
A6402-25MG04061832269054
A6402-5MG04061832674346