Skip to Content
Merck
All Photos(3)

Documents

A6376

Sigma-Aldrich

N-Acetyl-L-tryptophan

≥99% (TLC), suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

N-Acetyl-L-tryptophan,

Assay

≥99% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C13H14N2O3/c1-8(16)15-12(13(17)18)6-9-7-14-11-5-3-2-4-10(9)11/h2-5,7,12,14H,6H2,1H3,(H,15,16)(H,17,18)/t12-/m0/s1

InChI key

DZTHIGRZJZPRDV-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

N-Acetyl-L-tryptophan (NAT, Ac-Trp-OH) is used a substance P NK1 tachykinin receptor antagonist. N-Acetyl-L-tryptophan is also used as a competitive inhibitor to identify, differentiate and characterized tryptophanase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James J Donkin et al.
Journal of neurotrauma, 28(2), 217-224 (2010-12-24)
Previous studies have demonstrated that the compound N-acetyl-L-tryptophan (NAT) reduces brain edema and improves functional outcome following traumatic brain injury (TBI). In this study we examined whether this effect was mediated via the neurokinin-1 receptor, and whether there was an
M Lehnig et al.
Free radical research, 41(5), 523-535 (2007-04-25)
Tryptophan and melatonin are nitrated by peroxynitrite; tryptophan residues in proteins are susceptible to attack by reactive nitrogen species. Nitrated tryptophan might therefore be used as a biomarker for the involvement of reactive species derived from nitrogen oxide in a
Renée J Turner et al.
Brain research, 1393, 84-90 (2011-04-07)
Previous studies have suggested that substance P (SP) plays a critical role in the development of brain oedema and functional deficits following traumatic brain injury and that SP receptor antagonism may improve outcome. No studies have described such a role
Kate M Lewis et al.
Anti-cancer drugs, 24(4), 344-354 (2013-02-15)
Emend, an NK1 antagonist, and dexamethasone are used to treat complications associated with metastatic brain tumours and their treatment. It has been suggested that these agents exert anticancer effects apart from their current use. The effects of the NK1 antagonists
James J Donkin et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 29(8), 1388-1398 (2009-05-14)
Brain edema and swelling is a critical factor in the high mortality and morbidity associated with traumatic brain injury (TBI). Despite this, the mechanisms associated with its development are poorly understood and interventions have not changed in over 30 years.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service