Skip to Content
Merck
All Photos(1)

Key Documents

A6354

Sigma-Aldrich

Anti-Adiponectin antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Adiponectin Antibody, Anti-Adiponectin antibody produced in rabbit, Anti-Adipocyte Complement-Related Protein of 30 kDa

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

western blot (chemiluminescent): 0.5-1 μg/mL using using human plasma or placenta, or an extract of differentiated 3T3-L1 mouse cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADIPOQ(9370)
mouse ... Adipoq(11450)

General description

Adiponectin is secreted from adipose tissue for the regulation of glucose and fatty acid break down.
Anti-Adiponectin is produced in rabbit using a synthetic peptide corresponding to amino acid residues of human adiponectin. Adiponectin, also designated as adipocyte complement-related protein of 30 kDa (Acrp30), AdipoQ, apM1, Gelatin-binding protein 28 kDa (GBP28) is an adipocyte-specific protein that circulates in the blood stream, where it accounts for approximately 0.01% of all plasma proteins. Adiponectin belongs to the complement factor C1q family that belongs to the soluble defense collagen superfamily. Adiponectin (ADIPOQ) protein has a carboxyl-terminal globular domain and an amino-terminal collagen domain.
It is recommended to dilute the antibody in phosphate buffered saline containing 0.5% non-fat dry milk for background staining reduction.

Specificity

Rabbit Anti-Adiponectin antibody recognizes human and mouse adiponectin.

Immunogen

synthetic peptide corresponding to amino acid residues 225-244 of human adiponectin with N-terminal added cysteine, conjugated to KLH.

Application

Anti-Adiponectin antibody produced in rabbit has been used in western blotting and immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Rabbit Anti-Adiponectin antibody has been used for immunohistochemical and western blot applications.

Biochem/physiol Actions

Adipose tissue has important roles in energy storage, glucose homeostasis and fat metabolism.(10) Increased adiponectin levels are associated with type I diabetes mellitus, anorexia nervosa, weight reduction, and chronic renal failure. Adiponectin (ADIPOQ) affects lipid catabolism by increasing fatty acid oxidation in muscle, modulates glucose uptake, alters liver gluconeogenesis and enhances insulin-sensitivity in both muscle and liver. ADIPOQ has anti-atherosclerotic and anti-inflammatory properties. ADIPOQ has a role in physiological processes such as energy homeostasis and obesity. Low levels of adiponectin are also linked with hypertension and dyslipidemia. Mutation in the gene encoding it is linked with risk for endometrial cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Fernandes-Santos et al.
Nutrition (Burbank, Los Angeles County, Calif.), 25(7-8), 818-827 (2009-03-10)
We analyzed the effect of peroxisome proliferator-activated receptor (PPAR) agonists on adipose tissue morphology, adiponectin expression, and its relation to glucose and insulin levels in C57BL/6 mice fed a high-fat high-sucrose (HFHS) diet. Male C57BL/6 mice received one of five
P A Nidhina Haridas et al.
The Journal of clinical endocrinology and metabolism, 100(10), E1299-E1307 (2015-07-24)
Circulating ANGPTL8 has recently been used as a marker of insulin action. We studied expression and insulin regulation of ANGPTL8 and ANGPTL3 in vivo and in vitro. Expression of ANGPTL8 and ANGPTL3 was studied in 34 paired samples of human
Pan-PPAR agonist beneficial effects in overweight mice fed a high-fat high-sucrose diet
Fernandes-SC, et al.
Nutrition, 25(7), 818-827 (2009)
Deborah K Oh et al.
Diabetes, obesity & metabolism, 9(3), 282-289 (2007-03-30)
Adipose tissue is an active metabolic tissue that secretes multiple metabolically important proteins, known as adipokines. Adiponectin is an important adipokine because of its beneficial effects on glucose and lipid metabolism. Low levels of adiponectin are associated with disease states
Eicosapentaenoic acid (EPA) vs. Docosahexaenoic acid (DHA): Effects in epididymal white adipose tissue of mice fed a high-fructose diet
Bargut TCL, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 123, 14-24 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service