Skip to Content
Merck
All Photos(1)

Key Documents

A6017

Sigma-Aldrich

ω-Aminohexyl–Agarose

saline suspension

Synonym(s):

Omega-Aminohexyl-Agarose, 1,6-Diaminohexane–Agarose (N-linked), 6-Aminohexyl–Agarose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

plant

form

saline suspension

extent of labeling

≥5 μmol per mL

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino of 1,6-diaminohexane

matrix spacer

1 atom

capacity

≥5 mg/mL binding capacity (bovine serum albumin)

suitability

suitable for chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Hydrophobic ligands can serve as potentselective adsorbents. Both ω-aminoalkyl and alkyl agaroses can interact with the hydrophobic regions found in various proteins. The ω-aminoalkyl agarose product offered here consists of activated agarose with 1,6-diaminohexane covalently attached to one of its amine groups.

Application

ω-Aminohexyl–Agarose has been used in the conjugation of fatty acids with chain length from two carbons to 20 for fatty acyl beads pull-down experiments.
ω-aminohexyl–agarose has been used for coupling of serotype b-specific carbohydrate antigen (SbAg) of Actinobacillus actinomycetemcomitans Y4 for isolation of anti-SbAg antibodies using protein chromatography.

Biochem/physiol Actions

ω-aminoalkyl-agaroses are used in chromatography. In these, the protein is retained by lipophilic association between the hydrocarbon side chains on the agarose and hydrophobic pockets in the protein.

Physical form

Suspension in 0.5 M NaCl containing preservative

Other Notes

For R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mandjiny et al.
Journal of chromatography, 616(2), 189-195 (1993-07-02)
Histidine, a pseudobiospecific ligand, had been utilized to purify several proteins such as chymosin, acidic protease, carboxypeptidase Y and immunoglobulin G (IgG). A detailed study was undertaken to purify IgG on histidine coupled to aminohexyl Sepharose [A. El-Kak and M.
L G Martini et al.
Pharmaceutical research, 12(11), 1786-1790 (1995-11-01)
The purpose of this study was to investigate the influence of hydration characteristics on the in vitro release of 5-fluorouracil from a swellable matrix prepared using a novel triblock copolymer of poly(epsilon-caprolactone) and poly(oxyethylene). Matrices were prepared by dry compression
Y Ruttyn et al.
Journal of chromatography, 491(2), 299-308 (1989-07-21)
Human plasma was chromatographed under different experimental conditions on aminohexyl Sepharose. A strong retention of factor VIII/vWf and of factors VII, II, IX and X was observed. A satisfactory stabilization of eluted factor VIII clotting activity was obtained after addition
D Couchie et al.
The Biochemical journal, 199(2), 441-446 (1981-11-01)
Chromatography on hexyl-agarose resolved a partially purified cyclic GMP-activated phosphodiesterase from rat liver into two peaks of activity: the first was eluted with 0.5 M-KCl and was cyclic AMP-specific. The second was tightly bound to hexyl-agarose and was not eluted
Hydrophobic chromatography: use for purification of glycogen synthetase.
Shaltiel S and Er-El Z
Proceedings of the National Academy of Sciences of the USA, 70, 778-781 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service