Skip to Content
Merck
All Photos(4)

Key Documents

A6005

Sigma-Aldrich

Amygdalin

≥99% (HPLC), from apricot kernels

Synonym(s):

D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, D-Mandelonitrile-β-gentiobioside, Amygdaloside, Laetrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO11
CAS Number:
Molecular Weight:
457.43
Beilstein:
66856
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

apricot kernels

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

<11% water (Karl Fischer)

color

white

mp

223-226  °C

solubility

water: 50 mg/mL, clear to slightly hazy, colorless

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C20H27NO11/c21-6-10(9-4-2-1-3-5-9)30-20-18(28)16(26)14(24)12(32-20)8-29-19-17(27)15(25)13(23)11(7-22)31-19/h1-5,10-20,22-28H,7-8H2/t10?,11-,12-,13-,14-,15+,16+,17-,18-,19-,20-/m1/s1

InChI key

XUCIJNAGGSZNQT-SWRVSKMJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amygdalin, a cyanide containing glycoside, may be used as a substrate to identify, differentiate and characterize enzymes such as maltase(s), emulsin(s) and β-glucosidase(s).

Biochem/physiol Actions

Cyanogenic glycoside that is a component of bitter almonds and apricot pits. There is no scientific evidence that amygdalin itself is an effective anti-cancer agent. Recent studies using β−glucoside linked to a tumor-associated monoclonal antibody to release cyanide at the tumor cell has shown significant cytotoxicity.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Sánchez-Pérez et al.
Plant physiology, 158(4), 1916-1932 (2012-02-23)
Amygdalin is a cyanogenic diglucoside and constitutes the bitter component in bitter almond (Prunus dulcis). Amygdalin concentration increases in the course of fruit formation. The monoglucoside prunasin is the precursor of amygdalin. Prunasin may be degraded to hydrogen cyanide, glucose
Anne Vinther Morant et al.
Plant physiology, 147(3), 1072-1091 (2008-05-10)
Lotus japonicus accumulates the hydroxynitrile glucosides lotaustralin, linamarin, and rhodiocyanosides A and D. Upon tissue disruption, the hydroxynitrile glucosides are bioactivated by hydrolysis by specific beta-glucosidases. A mixture of two hydroxynitrile glucoside-cleaving beta-glucosidases was isolated from L. japonicus leaves and
Juan J Perez
Future medicinal chemistry, 5(7), 799-808 (2013-05-09)
Psoriasis is one of the most prevalent immune-mediated illness worldwide. The disease can still only be managed rather than cured, so treatments are aimed at clearing skin lesions and preventing their recurrence. Several treatments are available depending on the extent
Amygdalin (Laetrile) and veterinary medicine.
D W Macy
Journal of the American Veterinary Medical Association, 171(3), 284-286 (1977-08-01)
Li Liu et al.
Journal of ethnopharmacology, 139(2), 381-387 (2011-11-30)
Persicae Semen (Taoren) and Carthami Flos (Honghua) used in pair which is named as Taoren-Honghua (TH) herb pair has been used in traditional Chinese medicine (TCM) for promoting blood circulation to dissipate blood stasis for many years in China. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service