Skip to Content
Merck
All Photos(4)

Key Documents

A4106

Sigma-Aldrich

N-Acetyl-D-glucosamine

≥95% (HPLC)

Synonym(s):

2-Acetamido-2-deoxy-D-glucose, D-GlcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
Beilstein:
1727589
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

form

powder

color

beige

mp

211 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear to cloudy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8?/m1/s1

InChI key

OVRNDRQMDRJTHS-RTRLPJTCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetyl-D-glucosamine (D-GlcNAc) is a derivitized glucose monomer found in polymers of bacterial cell walls, chitin, hyaluronic acids and various glycans. D-GlcNAc is used to identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colette Cywes-Bentley et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), E2209-E2218 (2013-05-30)
Microbial capsular antigens are effective vaccines but are chemically and immunologically diverse, resulting in a major barrier to their use against multiple pathogens. A β-(1→6)-linked poly-N-acetyl-d-glucosamine (PNAG) surface capsule is synthesized by four proteins encoded in genetic loci designated intercellular
Lucy G Weaver et al.
Carbohydrate research, 371, 68-76 (2013-03-26)
Poly-N-acetylglucosamine (PNAG) saccharides are an important constituent of bacterial biofilms, such as those produced by Staphylococcus aureus. We have developed a simple two-step iterative method for the synthesis of β-(1→6)-glucosamine oligosaccharides that are structurally similar to PNAG. We illustrate the
Kirsi Rilla et al.
The Journal of biological chemistry, 288(8), 5973-5983 (2013-01-11)
Mammals have three homologous genes encoding proteins with hyaluronan synthase activity (Has1-3), all producing an identical polymer from UDP-N-acetylglucosamine and UDP-glucuronic acid. To compare the properties of these isoenzymes, COS-1 cells, with minor endogenous hyaluronan synthesis, were transfected with human
N-acetylglucosamine: a new osmotic solute in peritoneal dialysis solutions.
A Breborowicz et al.
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis, 17 Suppl 2, S80-S83 (1997-01-01)
Gerald W Hart et al.
Nature, 446(7139), 1017-1022 (2007-04-27)
All animals and plants dynamically attach and remove O-linked beta-N-acetylglucosamine (O-GlcNAc) at serine and threonine residues on myriad nuclear and cytoplasmic proteins. O-GlcNAc cycling, which is tightly regulated by the concerted actions of two highly conserved enzymes, serves as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service