Skip to Content
Merck

Skip To

A3672

Azurin

from Pseudomonas aeruginosa, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
235-551-8
NACRES:
NA.61
MDL number:
Form:
lyophilized powder
Biological source:
Pseudomonas aeruginosa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Pseudomonas aeruginosa

Quality Segment

form

lyophilized powder

composition

Protein, ≥65% Lowry

concentration

≥65.0% (Lowry)

technique(s)

toxicology assay: suitable

solubility

water: soluble 1—1.1 mg/mL, clear, blue (light blue to blue)

UniProt accession no.

storage temp.

−20°C

Gene Information

Pseudomonas aeruginosa ... AZU(878046)

General description

Research area: Apoptosis. Azurinis a periplasmic protein and is a homotetramer.[1]

Application

Azurin has been used:
  • in the cytotoxicity and cell viability studies in human osteosarcoma cell line
  • for the functionalization of silicon nitride cantilevers for interaction studies
  • for coating gold surface and insulating functionalized oxide surfaces of silicon oxide and mica

Biochem/physiol Actions

Azurin acts as an electron donor for nitrite reductase in bacterial denitrification process.[1] It exhibits anticancer activity[1] as it hampers various independent signaling pathways associated with cancer progression. It binds to tumor suppressor protein p53 and induces cancer cell apoptosis or stalls cancer cell growth. Azurin disrupts angiogenesis by reducing the activity of VEGFR-2tyrosine kinase thereby inhibiting tumor growth. It has been observed to show cytotoxicity in human breast cancer cells and human melanoma cells.[2]
Azurin is a metalloprotein in the family of cupredoxins. It preferentially enters cancer cells over normal cells and induces apoptosis. Azurin has structural similarities to ephrinB2, and in fact binds the ephrin receptor tyrosine kinase EphB2 to initiate cell signaling that is involved in cancer progression. Azurin inhibits autophosphorlyation of the EphB2 tyrosine residue, interfering with upstream cell signaling and contributing to cancer cell growth inhibition.

Physical form

Lyophilized powder containing ammonium acetate buffer salts.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6515F6303L9143
biological source

Pseudomonas aeruginosa

biological source

-

biological source

insect (firefly)

biological source

Pseudomonas aeruginosa (10)

technique(s)

toxicology assay: suitable

technique(s)

toxicology assay: suitable

technique(s)

toxicology assay: suitable

technique(s)

-

concentration

≥65.0% (Lowry)

concentration

-

concentration

-

concentration

-

form

lyophilized powder

form

powder

form

solid

form

lyophilized powder

Gene Information

Pseudomonas aeruginosa ... AZU(878046)

Gene Information

human ... CCK(885)
rat ... Cckbr(25706)

Gene Information

-

Gene Information

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. How do i know that this is azurin i mean any confirmatory test or anything else.

    1 answer
    1. The method of preparation is considered proprietary. However, as mentioned in this properties section of the Product Detail Page, this is Azurin from Pseudomonas aeruginosa. See the links below to review a sample Certificate of Analysis and the product data sheet, respectively:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/A3672-1MG0000211689.pdf
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/306/100/a3672dat.pdf

      Helpful?

Reviews

No rating value

Active Filters