Skip to Content
Merck
All Photos(4)

Key Documents

A3594

Sigma-Aldrich

ACES

BioPerformance Certified, ≥99.0%, suitable for cell culture

Synonym(s):

N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2NHCH2CH2SO3H
CAS Number:
Molecular Weight:
182.20
Beilstein:
2253770
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified

Quality Level

Assay

≥99.0%

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and bioburden, tested

pH

3.0-4.5

useful pH range

6.1-7.5

pKa (25 °C)

6.8

mp

>220 °C (dec.) (lit.)

solubility

water: 0.0527 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

NC(=O)CNCCS(O)(=O)=O

InChI

1S/C4H10N2O4S/c5-4(7)3-6-1-2-11(8,9)10/h6H,1-3H2,(H2,5,7)(H,8,9,10)

InChI key

DBXNUXBLKRLWFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Cu(II) complex formation by ACES buffer.: This study investigates the formation of copper(II) complexes with ACES buffer, providing insight into the biochemical interactions and stability of these complexes, which are significant for various biochemical applications. (Zawisza et al., 2013).

  • Differentiation of bacterial autolysins by zymogram analysis.: This research utilizes ACES buffer in zymogram analysis to differentiate bacterial autolysins, highlighting its effectiveness in maintaining enzyme activity and facilitating the study of bacterial cell wall degradation. (Strating and Clarke, 2001).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shadi Asadollahi et al.
International clinical psychopharmacology, 30(3), 142-150 (2014-12-17)
The objective of this study was to compare the efficacy of valproate versus haloperidol in decreasing the agitation level in affected patients in the emergency department. We assigned 80 acutely agitated patients to receive either intravenous sodium valproate (20 mg/kg)
P H Edelstein et al.
Journal of clinical microbiology, 31(12), 3329-3330 (1993-12-01)
Growth of Legionella spp. on buffered charcoal yeast extract medium supplemented with alpha-ketoglutarate and formulated with 3-(n-morpholino)propanesulfonic acid (MOPS), 3-(n-morpholino)-2-hydroxypropanesulfonic acid (MOPSO), or n-(2-acetamido)-2-aminoethanesulfonic acid (ACES) buffer was similar. With three exceptions, growth was no different in buffered yeast extract
A Alonso
Electrophoresis, 10(7), 513-519 (1989-07-01)
The polymorphism of alpha-1-antitrypsin (PI) has been studied by hybrid isoelectric focusing in miniaturized immobilized pH gradient gels, with an interelectrode distance of 55 mm, in two narrow ranges of pH 4.35-4.75 and 4.35-4.55), following rehydration with pH 4.2-4.9 carrier
D J Chappell
Clinical chemistry, 31(8), 1384-1386 (1985-08-01)
The hereditary deficiency variants of alpha 1-antitrypsin that are associated with diseases such as emphysema are usually identified by use of isoelectric focusing on polyacrylamide gels. Agarose is a simpler, faster, safer, and more reliable medium for this, but resolution
Free metal ion depletion by "Good's" buffers. II. N-(2-acetamido)-2-aminoethanesulfonic acid (ACESH): complexes with calcium(II), magnesium(II), manganese(II), cobalt(II), zinc(II), nickel(II), and copper(II).
J M Pope et al.
Analytical biochemistry, 103(2), 214-221 (1980-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service