Skip to Content
Merck

A3559

Albumin from mouse serum

lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)

Synonym(s):

Mouse albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
mouse
Assay:
≥99% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
immunohistochemistry: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

assay

≥99% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

immunohistochemistry: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

mouse ... ALB(11657)

General description

Albumin, present in the blood, is the most abundant protein. Structurally, albumin comprises three homologous domains (DI, DII, and DIII) with 67% α-helices.

Application

Albumin from mouse serum has been used:
  • as a standard in the enzyme-linked immunosorbent assay (ELISA) with urine samples
  • in the preparation of isolevuglandin (IsoLG)-albumin adducts for immunohistochemical staining of human tissue samples
  • for comparative studies with thymic stromal lymphopoietin (TSLP) to test its effect on dermal thickness in mice

Biochem/physiol Actions

Albumin transports fatty acids, drugs, and hormones. Mouse serum albumin (MSA) binds to the neonatal Fc receptor (FcRn).

Preparation Note

Prepared from albumin (A3139).

Other Notes

Albumin from mouse serum is free of globulin. It is derived from Swiss Webster strain mouse serum by cold alcohol fractionation. There are no heating steps involved in the preparation of this product.
This Fraction V albumin can be used for making immune complexes with anti-albumin antibodies. Fatty acids will not interfere with the complex formation.


Still not finding the right product?

Explore all of our products under Albumin from mouse serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library