Skip to Content
Merck

Skip To

A3048

Ammonium phosphate monobasic

suitable for plant cell culture

Synonym(s):

mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
NACRES:
NA.71
PubChem Substance ID:
UNSPSC Code:
12352207
EC Number:
231-764-5
MDL number:
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

technique(s)

cell culture | plant: suitable

mp

190 °C (dec.) (lit.)

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8843109709216003
form

powder

form

-

form

-

form

crystals or chunks

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

mp

190 °C (dec.) (lit.)

mp

190 °C (dec.) (lit.)

mp

190 °C (dec.) (lit.)

mp

190 °C (dec.) (lit.)

technique(s)

cell culture | plant: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, titration: suitable

technique(s)

-

technique(s)

-


Still not finding the right product?

Explore all of our products under Ammonium phosphate monobasic


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters