Skip to Content
Merck
All Photos(1)

Key Documents

A2979

Sigma-Aldrich

A-317491 sodium salt hydrate

≥98% (HPLC), powder

Synonym(s):

5-[[[(3-Phenoxyphenyl)methyl][(1S)-1,2,3,4-tetrahydro-1-naphthalenyl]amino]carbonyl]-1,2,4-benzenetricarboxylic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H27NO8 · xNa+ · yH2O
Molecular Weight:
565.57 (anhydrous free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL, clear

originator

Abbott

SMILES string

[Na].[H]O[H].OC(=O)c1cc(C(O)=O)c(cc1C(O)=O)C(=O)N(Cc2cccc(Oc3ccccc3)c2)[C@H]4CCCc5ccccc45

InChI

1S/C33H27NO8.Na.H2O.H/c35-30(25-17-27(32(38)39)28(33(40)41)18-26(25)31(36)37)34(29-15-7-10-21-9-4-5-14-24(21)29)19-20-8-6-13-23(16-20)42-22-11-2-1-3-12-22;;;/h1-6,8-9,11-14,16-18,29H,7,10,15,19H2,(H,36,37)(H,38,39)(H,40,41);;1H2;/t29-;;;/m0.../s1

InChI key

VEMCGHRDAPMXLI-VYEUTZPBSA-N

Application

A-317491 sodium salt hydrate has been used to block the P2X3 receptor. It has also been used to block the increased membrane conductance.

Biochem/physiol Actions

A-317491 helps to decrease neuropathic and inflammatory pain by inhibiting the P2X3 receptor-mediated Ca2+ influx.
Novel P2X3 and P2X2/3 receptor antagonist

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Subject to U.S. Patent No. 6,831,193. Sold under license from Abbott Laboratories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of A-317491 delivered by glycolipid-like polymer micelles on endometriosis pain
Yuan M, et al.
International journal of nanomedicine, 12, 8171-8171 (2017)
Early life allergen-induced mucus overproduction requires augmented neural stimulation of pulmonary neuroendocrine cell secretion
Barrios J, et al.
Faseb Journal, 31(9), 4117-4128 (2017)
Michael F Jarvis et al.
The Journal of pharmacology and experimental therapeutics, 310(1), 407-416 (2004-03-17)
A-317491 is a potent and selective antagonist of P2X3 and P2X(2/3) receptors. In the present studies, the ability of [3H]A-317491 to label recombinant human P2X(2/3) and P2X(3) receptors was characterized. Using membranes prepared from 1321N1 cells expressing P2X(2/3) receptors, [3H]A-317491
Michael F Jarvis et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(26), 17179-17184 (2002-12-17)
P2X3 and P2X2/3 receptors are highly localized on peripheral and central processes of sensory afferent nerves, and activation of these channels contributes to the pronociceptive effects of ATP. A-317491 is a novel non-nucleotide antagonist of P2X3 and P2X2/3 receptor activation.
Electrophysiological Evidence for Functional Astrocytic P2X3 Receptors in the Mouse Trigeminal Caudal Nucleus
Lee J, et al.
Experimental Neurobiology, 27(2), 88-93 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service