Product Name
[Val5]-Angiotensin II acetate salt hydrate, ≥95% (HPLC), powder
Quality Level
assay
≥95% (HPLC)
form
powder
composition
acetate, 1.2-10.4%
storage temp.
−20°C
SMILES string
O.CC(O)=O.CC(C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CC(O)=O)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc2cnc[nH]2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O
InChI
1S/C49H69N13O12.C2H4O2.H2O/c1-26(2)39(60-42(67)33(12-8-18-54-49(51)52)56-41(66)32(50)23-38(64)65)45(70)57-34(20-29-14-16-31(63)17-15-29)43(68)61-40(27(3)4)46(71)58-35(22-30-24-53-25-55-30)47(72)62-19-9-13-37(62)44(69)59-36(48(73)74)21-28-10-6-5-7-11-28;1-2(3)4;/h5-7,10-11,14-17,24-27,32-37,39-40,63H,8-9,12-13,18-23,50H2,1-4H3,(H,53,55)(H,56,66)(H,57,70)(H,58,71)(H,59,69)(H,60,67)(H,61,68)(H,64,65)(H,73,74)(H4,51,52,54);1H3,(H,3,4);1H2/t32-,33-,34-,35-,36-,37-,39-,40-;;/m0../s1
InChI key
INQPQDZJLFYCRH-AXOAQIBVSA-N
Gene Information
human ... AGTR1(185)
mouse ... AGTR1(11607)
rat ... AGTR1(24180)
General description
Application
- renal damage in mice to study the role of human liver-type fatty acid-binding protein (hL-FABP)
- vascular inflammation in mice to study the interaction of cytosolic copper chaperone antioxidant-1 (Atox1) with tumor necrosis factor (TNF)-receptor-associated factor 4 (TRAF4)
- abdominal aortic aneurysm (AAA) in mice to study the role of vascular ATPase copper transporting α (ATP7α)
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A2900-10MG | 04061832268972 |
| A2900-50MG | 04061832674209 |
| A2900-1MG | 04061823010900 |
| A2900-5MG | 04061832674223 |