Skip to Content
Merck

Skip To

A2478

APHA Compound 8

solid, ≥98% (HPLC)

Synonym(s):

3-(1-Methyl-4-phenylacetyl-1H-2-pyrrolyl)-N-hydroxy-2-propenamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H16N2O3
CAS Number:
Molecular Weight:
284.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

APHA Compound 8, ≥98% (HPLC), solid

Quality Segment

assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL, H2O: insoluble

storage temp.

2-8°C

SMILES string

Cn1cc(cc1\C=C\C(=O)NO)C(=O)Cc2ccccc2

InChI

1S/C16H16N2O3/c1-18-11-13(10-14(18)7-8-16(20)17-21)15(19)9-12-5-3-2-4-6-12/h2-8,10-11,21H,9H2,1H3,(H,17,20)/b8-7+

InChI key

UFQOXIMRSMFQRI-BQYQJAHWSA-N

Biochem/physiol Actions

HDAC (histone deacetylase) inhibitor.
One of a class of aroyl pyrrole hydroxy amide (APHA) compounds showing histone deacetylase (HDAC) inhibition, Compound 8 is the most potent and HDAC class I-selective.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3060A9227S5192
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

solid

form

powder

form

solid

form

solid

solubility

DMSO: >10 mg/mL, H2O: insoluble

solubility

DMSO: 2 mg/mL, clear

solubility

H2O: freely soluble 22 mg/mL

solubility

DMSO: soluble ~28 mg/mL, H2O: insoluble

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters