Skip to Content
Merck

A2202

ω-Agatoxin TK

~98% (HPLC), powder

Synonym(s):

ω-Aga-TK

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C215H337N65O70S10
CAS Number:
Molecular Weight:
5273.02
UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Form:
powder
Assay:
~98% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~98% (HPLC)

Quality Segment

form

powder

solubility

H2O: soluble (Aqueous stock solution is stable for 1 month at 2-8 °C and for several months at -20 °C.)

storage temp.

−20°C

Gene Information

human ... CACNA1A(773)
mouse ... CACNA1A(12286)
rat ... CACNA1A(25398)

Biochem/physiol Actions

Toxin found in the venom of the funnel web spider, Agelenopsis aptera. Potent, selective P-type and Q-type calcium-channel blocker, without effect on L-type, N-type, or T-type calcium channels. Used to differentiate the types of calcium channel active during various physiological processes


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library