Skip to Content
Merck
All Photos(1)

Key Documents

A2054

Sigma-Aldrich

Anti-Monkey IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Monkey IgG (whole molecule)-HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

monkey

technique(s)

direct ELISA: 1:45,000
dot blot: 1:160,000 (direct chemiluminescence assay)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulins (Igs) have two heavy (H) and two light (L) chains, held together by disulphide linkages. They belong to the immunoglobulin super-family. The heavy chain has one variable N-terminal region and three to four constant (CH1-CH4) C-terminal regions. The light chain has one variable and constant region. IgGs are subclassified into IgG1, IgG2, IgG3 and IgG4.
Monkey IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in monkey serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Horseradish Peroxidase (HRP) is an enzyme that catalyzes the conversion of chromogenic substrates such as o-phenylenediamine (OPD), 4-chloro-1-naphthol 3,3′,5,5′-tetramethylbenzidine (TMB), 3,3′-Diaminobenzidine (DAB) or 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) (ABTS); chemiluminescent substrates such as CPS-3 (enhanced luminal) and fluorogenic substrates such as Ampliflu Red into detectable chromophores, light-emitters or fluorescers, respectively.
Rabbit polyclonal anti-Monkey IgG (whole molecule)−Peroxidase antibody recognizes monkey IgG in vitro and in monkey serum and biological fluids.

Immunogen

monkey IgG (rhesus)

Application

Anti-Monkey IgG (whole molecule)−Peroxidase antibody produced in rabbit has been used in ELISA of serum samples.
Rabbit polyclonal anti-Monkey IgG (whole molecule)−Peroxidase antibody may be used to detect and quantitate the level of IgG in monkey serum and biological fluids via chromogenic, chemoluminescent or fluorogenic immunochemical or immunohistochemical techniques. It may also be used as a secondary antibody in assays that use monkey IgG as the primary antibody. It may be use to detect parasite-specific antibodies in monkey sera, ELISA assays were performed using HRP-conjugated rabbit anti-monkey IgG as the secondary.[1]

Biochem/physiol Actions

Digestion of IgG by papain results in generation of fragment antigen binding (Fab). Pepsin digestion of IgG results in fragment crystallizable (Fc). Fc regions of IgG antibody have enormous therapeutic potential and are exploited for the development of therapeutic antibodies. IgG1 class is the most abundant and its deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions and IgG4 is associated with asymptomatic infection. The IgG levels in monkey show age related increase. In monkeys, IgG subtypes comprises the predominant antibodies during immune response after hepatitis B surface antigen (HBsAg) vaccination. The human and monkey IgG sequences show divergence only in the hinge region and hence exploited for therapeutic antibody testing.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison and characterization of immunoglobulin G subclasses among primate species
Shearer MH, et al.
Clinical and Diagnostic Laboratory Immunology, 6(6), 953-958 (1999)
An attenuated Lassa vaccine in SIV-infected rhesus macaques does not persist or cause arenavirus disease but does elicit Lassa virus-specific immunity
Zapata JC, et al.
Virology Journal, 10(1), 52-52 (2013)
Nerea Zabaleta et al.
Cell host & microbe, 29(9), 1437-1453 (2021-08-25)
The SARS-CoV-2 pandemic has affected more than 185 million people worldwide resulting in over 4 million deaths. To contain the pandemic, there is a continued need for safe vaccines that provide durable protection at low and scalable doses and can
Vincentius Arca Testamenti et al.
Veterinary sciences, 7(4) (2020-10-15)
Burkholderia pseudomallei, the Gram-negative bacterium which causes melioidosis, is a threat to human and a wide range of animal species. There is an increased concern of melioidosis in Indonesian primate facilities, especially following case reports of fatal melioidosis in captive
Renato Porrozzi et al.
The American journal of tropical medicine and hygiene, 71(3), 297-305 (2004-09-24)
This study evaluates cross-immunity in rhesus monkeys (Macaca mulatta) previously infected with one species of Leishmania and have had self-cured disease or were cured by antimony-based therapy upon development of full-blown disease. We found that a self-healing cutaneous leishmaniasis (CL)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service