Skip to Content
Merck
All Photos(1)

Documents

A2024

Sigma-Aldrich

Apramycin sulfate salt

Synonym(s):

Nebramycin II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H41N5O11 · xH2SO4
CAS Number:
Molecular Weight:
539.58 (free base basis)
EC Number:
MDL number:
UNSPSC Code:
51281633
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces tenebrarius

form

powder

color

white to brown

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]4O[C@H](CO)[C@@H](N)[C@H](O)[C@H]4O

InChI

1S/C21H41N5O11.H2O4S/c1-26-11-14(30)18-8(33-20(11)37-21-16(32)13(29)10(25)9(4-27)34-21)3-7(24)19(36-18)35-17-6(23)2-5(22)12(28)15(17)31;1-5(2,3)4/h5-21,26-32H,2-4,22-25H2,1H3;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11+,12+,13+,14-,15-,16-,17-,18+,19+,20-,21-;/m1./s1

InChI key

WGLYHYWDYPSNPF-RQFIXDHTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Apramycin is used to study antibiotic resistance as well as protein synthesis translocation-step inhibition in bacteria and prokaryotes.

Biochem/physiol Actions

Apramycin inhibits protein synthesis by blocking translocation. It is also able to bind to the eukaryotic decoding site. At low concentrations it inhibits elongation and induces misreading of mRNA during protein synthesis.

Packaging

1G, 5G

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.Storage class (TRGS 510): Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T-W Hahn et al.
Journal of animal science, 84(6), 1422-1428 (2006-05-16)
Two experiments were conducted to evaluate the efficacy of beta-glucan on growth performance, nutrient digestibility, and immunity in weanling pigs. In Exp. 1, 210 weanling pigs (6.38 +/- 0.92 kg of BW) were fed dietary beta-glucan (0, 0.01, 0.02, 0.03
Min-Jung Choi et al.
Foodborne pathogens and disease, 8(1), 119-123 (2011-01-11)
A total of 1921 Escherichia coli isolated from healthy animals (501 from cattle, 832 from pigs, and 588 from chickens) and 237 isolates from diseased pigs were tested to determine the prevalence of apramycin and gentamicin resistance in Korea during
C M Yates et al.
The Journal of antimicrobial chemotherapy, 54(2), 534-537 (2004-07-03)
The aminoglycoside apramycin has been used extensively in animal husbandry in the UK since 1978. This study aimed to determine both whether calves that had never been treated with aminoglycoside antibiotics harboured apramycin-resistant (apr(R)) commensal Escherichia coli, and the mode
Kristina Kadlec et al.
The Journal of antimicrobial chemotherapy, 67(10), 2342-2345 (2012-06-22)
The aims of this study were to identify small staphylococcal plasmids that carry either the trimethoprim resistance gene dfrK or the apramycin resistance gene apmA and analyse them for their structure and organization with regard to their potential role as
Paulo Martins da Costa et al.
Veterinary microbiology, 139(3-4), 284-292 (2009-07-08)
The present study investigates, under field conditions, the influence of antimicrobial administration on prevalence and patterns of antimicrobial resistance among Escherichia coli and Enterococcus spp. isolated from growing broilers. For this purpose, a group of 16,000 commercial broiler chickens was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service