Skip to Content
Merck
All Photos(2)

Key Documents

A1960

Sigma-Aldrich

Aggrecan from bovine articular cartilage

lyophilized powder

Synonym(s):

Aggrecan Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

bovine articular cartilage

Quality Level

form

lyophilized powder

packaging

glass bottle of 1 mg

technique(s)

cell culture | mammalian: suitable

impurities

salt, essentially free

color

white

solubility

H2O: 2 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Application

Aggrecan is a critical component for cartilage structure and the function of joints. Its synthesis and degradation are being studied for their roles in cartilage deterioration during joint injury disease and aging.

Biochem/physiol Actions

This molecule produces a rigid, reversibly deformable gel that resists compression. It combines with hyaluronic acid to form very large macromolecular complexes that have an increased hydrohynamic volume and a significant increase (30-40%) in the relative viscosity of the solution.

Components

Aggrecan is a major structural proteoglycan of the cartilage extracellular matrix. It is a large proteoglycan, with a molecular weight greater than 2,500 kDa composed of approximately 100-150 glycosaminoglycan chains attached to a core protein. The majority of these glycosaminoglycan chains are chondroitin/dermatan sulfate. Aggrecan contains three globular domains, G1, G2, and G3, which are involved in aggregation and hylauronan binding, cell adhesion and chondrycte apoptosis.

Caution

Store this product at -20°C. Stored as supplied, this powder shows little decomposition in 3 years when stored properly.

Preparation Note

This product is extracted from articular cartilage, chematographically purified, dialyzed against water, and 0.2 μm filtered prior to lyophilization. Once lyophilized, this powder is essentially salt-free. The product is soluble in water at 2 mg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T E Hardingham et al.
European journal of clinical chemistry and clinical biochemistry : journal of the Forum of European Clinical Chemistry Societies, 32(4), 249-257 (1994-04-01)
Aggrecan, the large aggregating proteoglycan from cartilage contains chondroitin sulphate and keratan sulphate attached to a multidomain protein core. It aggregates by binding to hyaluronan and this is further stabilised by a separate globular link protein. There are two structurally
N Ishiguro et al.
Arthritis and rheumatism, 42(1), 129-136 (1999-01-27)
To determine the relationship between matrix metalloproteinases (MMPs), their inhibitors, and the turnover of matrix molecules in articular cartilage from patients with osteoarthritis (OA). Synovial fluid samples were collected from the knees of 54 patients with OA. Radiographic evaluations and
L Cao et al.
Experimental cell research, 246(2), 527-537 (1999-02-02)
A major feature of cartilage deterioration during joint injury and disease is aggrecan degradation and the loss of proteoglycan. Most of the degraded fragments are released into the circulatory system except the G1 domain which accumulates locally in the synovial
Doege, K.J. et al.
Guidebook to the Extracellular Matrix and Adhesion Proteins, 17-18 (1993)
C J Billington et al.
The Biochemical journal, 336 ( Pt 1), 207-212 (1998-11-10)
The breakdown of aggrecan in cartilage is, in part, mediated by an enzyme named aggrecanase that cleaves within the interglobular domain of the molecule between a glutamic residue and an alanine residue. Although the enzyme cleavage site has been identified

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service