A11629L2
optigel Precast gels
15-20% polyacrylamide, contains bind silane (long plate), with 2 orientation markers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
Quality Level
shelf life
1 mo.
contains
bind silane (long plate)
feature
with 2 orientation markers
concentration
in 0.375 M Tris-HCl, pH 8.8
15-20% polyacrylamide
gel L × W × thickness
256 mm × 200 mm × 1 mm
plate L × W × thickness
276 mm × 211 mm × 3 mm
276 mm × 217 mm × 3 mm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Optigels offer a range of large format pre-cast polyacrylamide gels ideal for use as the second dimension in 2D gel electrophoresis. Optigels are supplied using low fluorescing glass to allow compatibility with both traditional and fluorescent labeling systems, and are available prepared with bind silane if required. Optigels are compatible with several large format electrophoresis units including Cytiva′s Ettan™ DALTtwelve, Ettan DALTsix and BioRad Protean® Plus. All gels are cast with Laemmli (Tris-HCl) chemistry without SDS, providing flexibility of application. Developed for customers who require ultimate separation performance and reproducibility, our automated casting process provides ultimate gel-to-gel and batch-to-batch reproducibility for a broad range of homogeneous and gradient gels.
Legal Information
Ettan is a trademark of Cytiva
PROTEAN is a registered trademark of Bio-Rad Laboratories, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service