Skip to Content
Merck
All Photos(3)

Key Documents

A0554

Sigma-Aldrich

4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium salt hydrate

powder, ≥80%

Synonym(s):

Disodium 4-acetamido-4′-isothiocyanato-stilben-2,2′-disulfonate, SITS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12N2Na2O7S3 · xH2O
CAS Number:
Molecular Weight:
498.46 (anhydrous basis)
Beilstein:
9177451
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium salt hydrate, ≥80%

Assay

≥80%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].CC(=O)Nc1ccc(\C=C\c2ccc(cc2S([O-])(=O)=O)N=C=S)c(c1)S([O-])(=O)=O

InChI

1S/C17H14N2O7S3.2Na/c1-11(20)19-15-7-5-13(17(9-15)29(24,25)26)3-2-12-4-6-14(18-10-27)8-16(12)28(21,22)23;;/h2-9H,1H3,(H,19,20)(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b3-2+;;

InChI key

NPAWAMRXPHRVQY-WTVBWJGASA-L

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent marker for membranes; inhibits anion permeability of membranes. Also inactivates Na,K-ATPase, by covalent modification of a specific Lys residue.

Other Notes

Predominantly trans-isomer

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis Kolosov et al.
Insect biochemistry and molecular biology, 116, 103263-103263 (2019-11-05)
Malpighian tubules (MTs) and the hindgut together constitute the excretory system of insects. Larvae of lepidopterans (butterflies and moths) demonstrate the so-called cryptonephric arrangement, where the distal blind end of each MT is embedded into the rectal complex. The rest
M Montana Quick et al.
Journal of the American Society for Mass Spectrometry, 28(7), 1462-1472 (2017-03-21)
N-terminal derivatization of peptides with the chromogenic reagent 4-acetamido-4-isothiocyanatostilbene-2,2-disulfonic acid (SITS) is demonstrated to enhance the efficiency of 266 nm ultraviolet photodissociation (UVPD). Attachment of the chromophore results in a mass shift of 454 Da and provides significant gains in the number
C H Pedemonte et al.
FEBS letters, 314(1), 97-100 (1992-12-07)
The sodium pump or Na,K-ATPase, maintains the Na+ and K+ gradients across eukaryotic cell membranes at the expense of ATP. Incubation of purified canine renal Na,K-ATPase with 4-acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (SITS) inhibited the ATPase activity. Both the labeling of the protein
Kai Zhao et al.
Cellular and molecular life sciences : CMLS, 67(15), 2665-2678 (2010-04-03)
We have reported that chymotrypsin B (CtrB) is not just a digestive enzyme but is also stored in lysosomes. Herein, we demonstrated a broad distribution of CtrB and explored the involvement of CtrB in apoptosis. Exposure of RH-35 cells to
Effect of two inhibitors of anion transport on the hydrolysis of glucose 6-phosphate by rat liver microsomes. Covalent modification of the glucose 6-P transport component.
M A Zoccoli et al.
The Journal of biological chemistry, 255(3), 1113-1119 (1980-02-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service