Skip to Content
Merck
All Photos(1)

Documents

94158

Sigma-Aldrich

Tris Buffered Saline

tablet (for 500 mL), pH 7.6, BioUltra

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

product name

Tris Buffered Saline, BioUltra, tablet (for 500 mL), pH 7.6

product line

BioUltra

Quality Level

form

tablet (for 500 mL)

pH

7.6

solubility

water: 2 tablet/liter, clear, colorless

application(s)

diagnostic assay manufacturing

Looking for similar products? Visit Product Comparison Guide

Application

Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris Buffered Saline, pH 7.6 may be used as a sample diluent, phase separation media or placebo.

Reconstitution

1 tablet dissolved in 500 mL of deionized water yields:
0.15 M sodium chloride; 0.050 M TRIS-HCl buffer; pH 7.6 at 25°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan C Arciniega et al.
Experimental eye research, 93(4), 452-459 (2011-07-02)
The purpose of this study was to evaluate the impact of free fatty acids (FFA), namely oleic (OA) and linoleic (LA) ones, on meibomian lipid films (MLF) using a Langmuir trough (LT) and a Brewster angle microscope (BAM). Human meibum
Charles L Balzli et al.
Current eye research, 35(6), 480-486 (2010-05-15)
Staphylococcus aureus is an important cause of ocular infections including endophthalmitis. The purpose of this study was to determine the ability of a relatively large molecule, such as lysostaphin, to remain in the rabbit aqueous humor for extended periods while
Christopher Kaufmann et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 98(2), 280-289 (2011-05-24)
Commercially available drug-eluting stents have the potential to induce inflammatory and hypersensitive adverse reactions due to their polymer coating. The use of self assembled monolayers (SAMs) as an alternate drug delivery platform for stents has recently been demonstrated. In this
Susanna Trapp et al.
Journal of virology, 83(2), 884-895 (2008-11-14)
Human immunodeficiency virus (HIV) is taken up by and replicates in immature dendritic cells (imDCs), which can then transfer virus to T cells, amplifying the infection. Strategies known to boost DC function were tested for their ability to overcome this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service