Skip to Content
Merck
All Photos(1)

Key Documents

94129

Sigma-Aldrich

Immobead 150P

0.15-0.50 mm particle size

Synonym(s):

Copolymer of methacrylate, cross-linked and carrying oxirane groups

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

beads

loss

≤10% loss on drying

particle size

0.15-0.50 mm

storage temp.

−20°C

Application

Immobead 150 0.15-0.30 mm particle size, a resin for enzyme/protein immobilization, is used for covalent binding of enzymes such as lipases. Bound enzymes are easy to repurify and use in industrial applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Ming Zheng et al.
Bioresource technology, 115, 141-146 (2012-01-03)
In this work, mixed-mode silica particles functionalized with octyl and sulfonic acid groups was conveniently prepared by co-bonding a mixture of n-octyltriethoxysilane and 3-mercaptopropyltriethoxysilane and then oxidized with hydrogen peroxide. Candida rugosa lipase (CRL) was immobilized on the mixed-mode silica
Bo Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 24(23), 13457-13464 (2008-11-11)
A series of epoxy-activated polymer films composed of poly(glycidyl methacrylate/butyl methacrylate/hydroxyethyl methacrylate) were prepared. Variation in comonomer composition allowed exploration of relationships between surface wettability and Candida antartica lipase B (CALB) binding to surfaces. By changing solvents and polymer concentrations
Lili Li et al.
Colloids and surfaces. B, Biointerfaces, 102, 124-129 (2012-09-27)
Amino-cyclodextrin was covalently attached to multiwalled carbon nanotubes (MWNTs). The functionalized MWNTs have a good dispersibility in water. The lipase was adsorbed onto the functionalized MWNTs. The immobilized lipase was utilized for the resolution of the model compound (R, S)-1-phenyl
Lipase Immobilization on Hypercrosslinked Polymer Coated Silica for Biocatalytic Synthesis of Phytosterol Esters with Controllable Fatty Acid Composition.
Zheng M, Lu Y, Huang F, Wang L, Guo P, et al.
Journal of Agricultural and Food Chemistry (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service