Skip to Content
Merck
All Photos(1)

Key Documents

92034

Sigma-Aldrich

sn-Glycerol 1-phosphate lithium salt

≥95.0% (TLC)

Synonym(s):

D-Glycerol 3-phosphate lithium salt, L-Glycerol 1-phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9O6P · xLi+
Molecular Weight:
172.07 (free acid basis)
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95.0% (TLC)

form

powder

optical activity

[α]/D -2.0±0.5°, c = 1 in H2O

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

OC[C@H](O)COP(O)(O)=O

InChI

1S/C3H9O6P/c4-1-3(5)2-9-10(6,7)8/h3-5H,1-2H2,(H2,6,7,8)/t3-/m0/s1

InChI key

AWUCVROLDVIAJX-VKHMYHEASA-N

Biochem/physiol Actions

Metabolite of glycerophospholipid metabolism, component of lipoteichoic acids and polyglycerolphosphates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosuke Koga et al.
Bioscience, biotechnology, and biochemistry, 70(1), 282-285 (2006-01-24)
sn-Glycerol-1-phosphate dehydrogenase is responsible for the formation of sn-glycerol-1-phosphate, the backbone of membrane phospholipids of Archaea. This activity had never been detected in cell-free extract of Sulfolobus sp. Here we report the detection of this activity on the thermostable ST0344
Harald Guldan et al.
Biochemistry, 47(28), 7376-7384 (2008-06-19)
The exclusive presence of glycerol-1-phosphate dehydrogenases (G1PDH) has been postulated to be a key feature that distinguishes archaea from bacteria. However, homologues of G1PDH genes can be found in several bacterial species, among them the hitherto uncharacterized open reading frame
Yan Boucher et al.
Molecular microbiology, 52(2), 515-527 (2004-04-07)
A characteristic feature of the domain archaea are the lipids forming the hydrophobic core of their cell membrane. These unique lipids are composed of isoprenoid side-chains stereospecifically ether linked to sn-glycerol-1-phosphate. Recently, considerable progress has been made in characterizing the
M Nishihara et al.
Journal of biochemistry, 117(5), 933-935 (1995-05-01)
One of the most characteristic features of archaebacterial ether phospholipids is the enantiomeric configuration of their glycerophosphate backbone (sn-glycerol-1-phosphate), that is the mirror image of the structure of the eubacterial or eukaryotic counterpart. The enzyme that forms glycerophosphate of this
Angelika Gründling et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(20), 8478-8483 (2007-05-08)
Lipoteichoic acid (LTA), a glycerol phosphate surface polymer, is a component of the envelope of Gram-positive bacteria. However, the molecular basis for its synthesis or function is not known. Here we report that Staphylococcus aureus LtaS synthesizes glycerol phosphate LTA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service