Skip to Content
Merck
All Photos(1)

Key Documents

88635

Sigma-Aldrich

5-Thio-D-glucose

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5S
CAS Number:
Molecular Weight:
196.22
Beilstein:
1865193
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D 220.0±5.0°, c = 1 in 0.1 M HCl

color

white to off-white

mp

135-138 °C (lit.)

SMILES string

OC[C@H]1S[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O5S/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

KNWYARBAEIMVMZ-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Thio-D-glucose (Glc-5S), a thiosugar hexokinase inhibitor, is used to control/reduce the rate of glycolysis in vivo.

Biochem/physiol Actions

Potent, competitive inhibitor of the cellular transport of D-glucose and of D-glucose mediated insulin release. Inhibitor of spermatogenesis.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyin Wu et al.
Journal of neurophysiology, 91(4), 1734-1747 (2003-12-03)
Circulating glucose levels significantly affect vagal neural activity, which is important in the regulation of pancreatic functions. Little is known about the mechanisms involved. This study investigates the neural pathways responsible for hypoglycemia-induced vagal efferent signaling to the pancreas and
Studies on oxidative phosphorylation. XVI. Sulfhydryl involvement in the energy-transfer pathway.
C K Kurup et al.
Biochemistry, 7(12), 4483-4491 (1968-12-01)
The pancreatic beta-cell recognition of insulin secretagogues. 3. Effects of substituting sulphur for oxygen in the D-glucose molecule.
B Hellman et al.
Biochemical pharmacology, 22(1), 29-35 (1973-01-01)
I Tahirovic et al.
Journal of neural transmission. Supplementum, (72)(72), 235-240 (2007-11-07)
It is believed that oxidative stress plays a central role in the pathogenesis of metabolic diseases like diabetes mellitus (DM) and its complications (like peripheral neuropathy) as well as in neurodegenerative disorders like sporadic Alzheimer's disease (sAD). Representative experimental models
S Ritter et al.
Brain research, 856(1-2), 37-47 (2000-03-10)
Feeding and blood glucose responses to local injection of nanoliter volumes of 5-thio-D-glucose (5TG), a potent antimetabolic glucose analogue, were studied at 142 hindbrain and 61 hypothalamic cannula sites. A site was considered positive if 5TG elicited at least 1.5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service