Skip to Content
Merck
All Photos(1)

Key Documents

86265

Sigma-Aldrich

D-(+)-Talose

≥99.0% (HPLC)

Synonym(s):

Aldehydo-D-talose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724617
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99.0% (HPLC)

form

crystals

optical activity

[α]20/D +21±1°, 3 hr, c = 1% in H2O

color

white

mp

124-127 °C

solubility

H2O: 0.1 g/mL

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-WHZQZERISA-N

Application

D-Talose, a C-2 epimer of galactose, is used as a substrate to identify, differentiate and characterize ribose-5-phosphate isomerase(s) of Clostridia.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enrico Ragni et al.
Cells, 9(5) (2020-05-14)
Extracellular vesicles (EVs) showed therapeutic properties in several applications, many in regenerative medicine. A clear example is in the treatment of osteoarthritis (OA), where adipose-derived mesenchymal stem cells (ASCs)-EVs were able to promote regeneration and reduce inflammation in both synovia
Yasuhiko Matsumoto et al.
PloS one, 6(3), e18292-e18292 (2011-04-12)
The number of individuals diagnosed with type 2 diabetes mellitus, which is caused by insulin resistance and/or abnormal insulin secretion, is increasing worldwide, creating a strong demand for the development of more effective anti-diabetic drugs. However, animal-based screening for anti-diabetic
Congai Han et al.
Nanomaterials (Basel, Switzerland), 10(4) (2020-03-29)
The reasonable design of magnetic carbon-based composites is of great significance to improving the microwave absorption (MA) performance of the absorber. In this work, ultrafine FeNi3 nanocrystals (5-7 nm) embedded in a 3D honeycomb-like carbon matrix (FeNi3@C) were synthesized via
Yuichiro Onishi et al.
Carbohydrate research, 493, 108031-108031 (2020-05-27)
Galactose was isomerized in pure water or in 10 mmol/L sodium phosphate buffer at 160 °C under pressurized conditions. The isomerization of galactose to tagatose and talose in phosphate buffer resulted in 14% and 1.4% yields, respectively, which were significantly higher than
Cun Juan Xia et al.
Materials (Basel, Switzerland), 13(19) (2020-09-30)
The microstructure evolution, tensile properties and work-hardening behavior of AA2219 alloy reinforced by in situ nanosized TiB2 particles were studied in this paper. The observation indicated an impeded recrystallization of the matrix alloy by nanosized TiB2 particles, and the hybrids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service