Skip to Content
Merck
All Photos(1)

Key Documents

82648

Sigma-Aldrich

Pyrene

puriss. p.a., for fluorescence, ≥99.0% (GC)

Synonym(s):

Benzo[def]phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1307225
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

grade

for fluorescence
puriss. p.a.

Quality Level

Assay

≥99.0% (GC)

form

crystals

mp

145-148 °C (lit.)
151-154 °C

solubility

ethanol: soluble

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

fluorescence

λex 338 nm; λem 375 nm in DMSO

SMILES string

c1cc2ccc3cccc4ccc(c1)c2c34

InChI

1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H

InChI key

BBEAQIROQSPTKN-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Analysis Note

In addition to the emission maximum at 375 nm, there are lower maxima at 395, 385, 380, 415 and 445 nm.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

435.2 °F

Flash Point(C)

224 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaowei Guo et al.
Nature communications, 11(1), 959-959 (2020-02-23)
Hydronium-ion batteries are regarded as one of the most promising energy technologies as next-generation power sources, benefiting from their cost effectivity and sustainability merits. Herein, we propose a hydronium-ion battery which is based on an organic pyrene-4,5,9,10-tetraone anode and an
Yan Fang et al.
Drug delivery, 23(9), 3582-3593 (2016-09-30)
A kind of polymeric lipid vesicles (PLVs) with pH-responsive turning on-off membrane for programed delivery of insulin in gastrointestinal (GI) tract was developed, which was self-assembled from the grafted amphipathic polymer of N-tocopheryl-N'-succinyl-ɛ-poly-l-lysine (TP/SC-g-PLL). By controlling the grafting ratio of
Intramolecular pyrene excimer fluorescence: a probe of proximity and protein conformational change.
S S Lehrer
Methods in enzymology, 278, 286-295 (1997-01-01)
Michael E Østergaard et al.
Chemical Society reviews, 40(12), 5771-5788 (2011-04-14)
Pyrene-functionalized oligonucleotides (PFOs) are increasingly explored as tools in fundamental research, diagnostics and nanotechnology. Their popularity is linked to the ability of pyrenes to function as polarity-sensitive and quenchable fluorophores, excimer-generating units, aromatic stacking moieties and nucleic acid duplex intercalators.
Pentti Somerharju
Chemistry and physics of lipids, 116(1-2), 57-74 (2002-07-03)
Pyrene is one of the most frequently used lipid-linked fluorophores. Its most characteristic features are a long excited state lifetime and (local) concentration-dependent formation of excimers. Pyrene is also hydrophobic and thus does not significantly distort the conformation of the

Related Content

Nancy-520 for DNA Detection and Quantitation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service