Skip to Content
Merck
All Photos(1)

Key Documents

79849

Sigma-Aldrich

(±)-Mevalonic acid 5-phosphate lithium salt hydrate

95% (TLC)

Synonym(s):

(±)-MVAP-Li, rac-5-Phosphomevalonate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O7P · xLi+ · yH2O
Molecular Weight:
228.14 (free acid basis)
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

95% (TLC)

form

powder or crystals

relevant disease(s)

Alzheimer′s disease; autoimmune diseases

storage temp.

2-8°C

SMILES string

OC(CC(C)(O)CCOP(O)(O)=O)=O.[H]O[H].[Li+]

InChI

1S/C6H13O7P.3Li.H2O/c1-6(9,4-5(7)8)2-3-13-14(10,11)12;;;;/h9H,2-4H2,1H3,(H,7,8)(H2,10,11,12);;;;1H2/q;3*+1;/p-3

InChI key

VYTYNAKRCDFACM-UHFFFAOYSA-K

Related Categories

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Gong et al.
Cell death & disease, 10(4), 327-327 (2019-04-13)
The mevalonate pathway is known for the synthesis of cholesterol, but recent studies have reported that it also controls Hippo signaling, which is critical for the regulation of organ size and tumorigenesis. Here, we discover that the suppression of the
D N Skilleter et al.
The Biochemical journal, 124(2), 407-417 (1971-09-01)
1. Phosphomevalonate kinase and 5-pyrophosphomevalonate decarboxylase have been purified from the freeze-dried latex serum of the commercial rubber tree Hevea brasiliensis. 2. The phosphomevalonate kinase was acid- and heat-labile and required the presence of a thiol to maintain activity. 3.
Inhibition of hepatic cholesterol biosynthesis by 3,5-dihydroxy-3,4,4-trimethylvaleric acid and its site of action.
F H Hulcher
Archives of biochemistry and biophysics, 146(2), 422-427 (1971-10-01)
John L Andreassi et al.
Biochemistry, 48(27), 6461-6468 (2009-06-03)
The galacto-, homoserine-, mevalonate-, phosphomevalonate-kinase (GHMP) superfamily encompases a wide-range of protein function. Three members of the family (mevalonate kinase, phosphomevalonate kinase, and diphosphomevalonate decarboxylase) comprise the mevalonate pathway found in S. pneumoniae and other organisms. We have determined the
K Audrey Audetat et al.
Molecular and cellular biology, 37(13) (2017-04-19)
The human Mediator complex regulates RNA polymerase II transcription genome-wide. A general factor that regulates Mediator function is the four-subunit kinase module, which contains either cyclin-dependent kinase 8 (CDK8) or CDK19. Whereas CDK8 is linked to specific signaling cascades and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service