Skip to Content
Merck
All Photos(1)

Key Documents

75025

Sigma-Aldrich

D-Erythronic acid potassium salt

≥97.0% (GC)

Synonym(s):

(2R,3R)-2,3,4-Trihydroxybutanoic acid potassium salt, Potassium D-erythronate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O5 · xK+
CAS Number:
Molecular Weight:
136.10 (free acid basis)
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97.0% (GC)

form

powder

optical activity

[α]/D 12.0±2.0°, c = 1 in H2O

color

white to off-white

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)C(O)=O.[K+]

InChI

1S/C4H8O5.K/c5-1-2(6)3(7)4(8)9;/h2-3,5-7H,1H2,(H,8,9);/q;+1/t2-,3-;/m1./s1

InChI key

NIBJBHRBFKKLFG-SWLXLVAMSA-N

Biochem/physiol Actions

Erythronic acid is a sugar component of aqueous humour (eye). Erythronic acid is a normal organic acid present in a healthy adult and pediatric population and has been found in urine, plasma, CSF, aqueous humour (eye) and synovial fluid. Erythronic acid is formed when GlcNAc is oxidised.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karsten Suhre et al.
PloS one, 5(11), e13953-e13953 (2010-11-19)
Metabolomics is the rapidly evolving field of the comprehensive measurement of ideally all endogenous metabolites in a biological fluid. However, no single analytic technique covers the entire spectrum of the human metabolome. Here we present results from a multiplatform study
Metabolomics of capsicum ripening reveals modification of the ethylene related-pathway and carbon metabolism.
Aizat, W.M., et al.
Postharvest biology and technology, 89, 19-31 (2014)
Noha A Yousri et al.
Metabolomics : Official journal of the Metabolomic Society, 10(5), 1005-1017 (2014-09-02)
Changes in an individual's human metabolic phenotype (metabotype) over time can be indicative of disorder-related modifications. Studies covering several months to a few years have shown that metabolic profiles are often specific for an individual. This "metabolic individuality" and detected
G F Hoffmann et al.
Journal of chromatography, 617(1), 1-10 (1993-07-23)
Quantitative reference values for the concentrations of organic acids in cerebrospinal fluid (CSF) and plasma, as well as ratios of individual organic acids between CSF and plasma, were determined in twenty-three pairs of samples from pediatric patients. Twenty-six organic acids
J J Harding et al.
Current eye research, 19(2), 131-136 (1999-07-27)
Sugars in the aqueous humour of the eye serve both as a source of nutrients to the lens and other anterior ocular tissues, and potentially as an indicator of waste products from these tissues. In this work we intended to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service