Skip to Content
Merck

74378

(2-Nonen-1-yl)succinic anhydride

suitable for electron microscopy

Synonym(s):

Dihydro-3-(nonen-1-yl)-2,5-furandione, Nonenylsuccinic anhydride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H20O3
CAS Number:
Molecular Weight:
224.30
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
249-317-8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

composition

(mixture of branched chain isomers)

technique(s)

electron microscopy: suitable

density

1.03 g/mL at 20 °C (lit.)

lipid type

unsaturated FAs

SMILES string

CCCCCC\C=C\CC1CC(=O)OC1=O

InChI

1S/C13H20O3/c1-2-3-4-5-6-7-8-9-11-10-12(14)16-13(11)15/h7-8,11H,2-6,9-10H2,1H3/b8-7+

InChI key

CIMXGTDNUHWJCZ-BQYQJAHWSA-N

Application

For use in Spurr embedding technique.


Still not finding the right product?

Explore all of our products under (2-Nonen-1-yl)succinic anhydride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

294.8 °F - closed cup

flash_point_c

146 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library