Skip to Content
Merck

72762

Blue Dextran

average mol wt 500,000

Synonym(s):

Blue Dextran 500, Dextran blue from Leuconostoc mesenteroides

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352116
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mol wt

average mol wt 500,000

loss

≤10% loss on drying

capacity

0.01-0.08 mmol/g loading

storage temp.

−20°C



Still not finding the right product?

Explore all of our products under Blue Dextran


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library