Skip to Content
Merck
All Photos(1)

Key Documents

69312

Sigma-Aldrich

L-Glyceraldehyde-3-phosphate solution

≥95% (TLC), 8-12 mg/mL in H2O (qNMR)

Synonym(s):

(2S)-2-Hydroxy-3-phosphonooxypropanal, L-GAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7O6P
CAS Number:
Molecular Weight:
170.06
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (TLC)

form

liquid

concentration

8-12 mg/mL in H2O (qNMR)

impurities

≤0.5 mg/mL D-glyceraldehyde-3-phosphate (enzymatic)

storage temp.

−20°C

SMILES string

[H]C([C@@H](O)COP(O)(O)=O)=O

InChI

1S/C3H7O6P/c4-1-3(5)2-9-10(6,7)8/h1,3,5H,2H2,(H2,6,7,8)/t3-/m1/s1

InChI key

LXJXRIRHZLFYRP-GSVOUGTGSA-N

Biochem/physiol Actions

L-Glyceraldehyde 3-phosphate is a potent non-competitive inhibitor of NADP-dependent non-phosphorylating D-glyceraldehyde-3-phosphate dehydrogenase and a substrate for L-Glyceraldehyde 3-phosphate reductase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C T Tang et al.
Antimicrobial agents and chemotherapy, 11(1), 147-153 (1977-01-01)
At a concentration of 2.5 mM, dl-glyceraldehyde 3-phosphate has a bactericidal effect upon Escherichia coli. The glycerol 3-phosphate transport system is required for the entry of the biologically active l-enantiomer. l-Glyceraldehyde must be phosphorylated by the cell to exert its
A Hall et al.
Biochemistry, 14(19), 4348-4353 (1975-09-23)
By a combination of methods involving enzyme-catalyzed reactions and classical iodination techniques it has been possible to obtain all the relevant rate constants for the uncatalyzed interconversion of dihydroxyacetone phosphate and D-glyceraldehyde 3-phosphate via their common enediol intermediate. These rate
Kevin K Desai et al.
Bioorganic chemistry, 38(1), 37-41 (2009-12-18)
Recently, we reported that YghZ from Escherichia coli functions as an efficient L-glyceraldehyde 3-phosphate reductase (Gpr). Here we show that Gpr co-purifies with a b-type heme cofactor. Gpr associates with heme in a 1:1 stoichiometry to form a complex that
Purification and properties of NADP-dependent non-phosphorylating glyceraldehyde-3-phosphate dehydrogenase from the green alga Chlamydomonas reinhardtii.
Iglesias, A.A., et al.
Biochimica et Biophysica Acta, 925, 1-10 (1987)
J P Richard
Biochemistry, 24(4), 949-953 (1985-02-12)
Triosephosphate isomerase catalyzes the isomerization and/or racemization reactions of L-glyceraldehyde 3-phosphate (LGAP), the enantiomer of the physiological substrate. The reaction is inhibited by the active site directed reagent glycidol phosphate. The amount of protonation product formation catalyzed by a fixed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service