Skip to Content
Merck
All Photos(1)

Documents

68795

Sigma-Aldrich

Hydrophobin SC3

≥98% (SDS-GE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

Quality Level

Assay

≥98% (SDS-GE)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

Schizophyllum commune H4-8 ... SC3(9592530)

General description

Hydrophobin SC3 is a class I hydrophobin of Schizophyllum commune. It is an oligomeric amphiphilic protein, rich in β-sheet. The mature protein consists of four disulfide bridges comprising eight cysteine residues. The N-terminal contains mannose residues in combination with threonine residues.

Application

Hydrophobin SC3 has been used to coat nitric oxide (NO) releasing medical-grade polymer to establish an antifouling layer to work synergistically with NO′s bactericidal and antiplatelet activity (SC3-NO).

Biochem/physiol Actions

Hydrophobins are key regulators of fungal growth and development. The hydrophobin SC3 is known for its ability to assemble at the water and air, water and oil, and water and a hydrophobic solid interface. The self-assembling process subjects hydrophobin SC3 to many conformational changes. It permits the growth of aerial hyphae and fruiting bodies on fungi as a result of assembling at the water-air interface. Hydrophobin SC3 mediates the linking of cell wall glucans to chitin and helps in the hyphal cell wall formation. Hydrophobin SC3 might have antitumor actions and be useful in chemotherapy and radiation.

Other Notes

Currently available only in Europe.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Wang et al.
Biophysical journal, 88(5), 3434-3443 (2005-03-08)
Hydrophobins are a class of small proteins that fulfill a wide spectrum of functions in fungal growth and development. They do so by self-assembling into an amphipathic membrane at hydrophilic-hydrophobic interfaces. The SC3 hydrophobin of Schizophyllum commune is the best-studied
Marijke Haas Jimoh Akanbi et al.
Applied microbiology and biotechnology, 97(10), 4385-4392 (2012-08-01)
The use of mushroom extracts has been common practice in traditional medicine for centuries, including the treatment of cancer. Proteins called hydrophobins are very abundant in mushrooms. Here, it was examined whether they have antitumor activity. Hydrophobin SC3 of Schizophyllum
Paul J Molino et al.
ACS nano, 12(11), 11610-11624 (2018-10-20)
Hydrophilic surface chemistries can strongly bind water to produce surfaces that are highly resistant to protein adsorption and fouling. The interfacial bound water and its distinct properties have intrigued researchers for decades, yet the relationship between the water three-dimensional structure
Ryan Devine et al.
Biomaterials science, 7(8), 3438-3449 (2019-07-04)
In medical device design, there is a vital need for a coating that promotes treatment of the patient and simultaneously prevents fouling by biomacromolecules which in turn can progress to infections, thrombosis, and other device-related complications. In this work, hydrophobin
Eva Sahakian et al.
Journal of leukocyte biology, 102(2), 475-486 (2017-05-28)
Epigenetic changes in chromatin structure have been recently associated with the deregulated expression of critical genes in normal and malignant processes. HDAC11, the newest member of the HDAC family of enzymes, functions as a negative regulator of IL-10 expression in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service