Skip to Content
Merck
All Photos(2)

Key Documents

62298

Sigma-Aldrich

Lipase from Mucor miehei

powder, slightly brown, ~1 U/mg

Synonym(s):

Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

Quality Level

specific activity

~1 U/mg

color

slightly brown

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Mucor miehei lipase hydrolizes triglycerides and catalyzes the synthesis of esters from glycerol and long-chain fatty acids in vitro. Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40 °C (triolein, Cat. No. 62314 as substrate)

Other Notes

Characterization; In the synthesis of fatty hydroxamic acids; Esterification

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Huge-Jensen et al.
Lipids, 22, 559-559 (1987)
P.A.S.S. Marques et al.
Journal of Chemical Technology and Biotechnology, 55, 25-25 (1992)
B. Huge-Jensen et al.
Journal of the American Oil Chemists' Society, 65, 905-905 (1988)
F. Servat et al.
Journal of the American Oil Chemists' Society, 67, 646-646 (1990)
J Hall et al.
The Journal of small animal practice, 54(5), 275-279 (2013-02-13)
Mushroom toxicosis is rarely diagnosed in dogs and is poorly reported in the veterinary literature. This report suggests that mushroom toxicosis is a potentially under-diagnosed condition in first opinion practice in the UK. Nine dogs with clinical signs consistent with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service