Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
MDL number:
UNSPSC Code:
51321705
EC Number:
232-307-2
NACRES:
NA.25
Beilstein/REAXYS Number:
5209585
biological source
egg yolk (dried)
Quality Segment
form
solid
concentration
≥50% (TLC)
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
ambient
storage temp.
−20°C
SMILES string
[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C
InChI
1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1
InChI key
JLPULHDHAOZNQI-ZTIMHPMXSA-N
Biochem/physiol Actions
A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.
Still not finding the right product?
Explore all of our products under L-α-Phosphatidylcholine
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.