Skip to Content
Merck
All Photos(1)

Documents

60528

Sigma-Aldrich

Potassium sulfate

BioUltra, ≥99.0% (AT)

Synonym(s):

Dipotassium sulfate, Potassium sulphate, Sulphate of potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

Quality Level

Assay

≥99.0% (AT)

form

solid

impurities

insoluble matter, passes filter test
≤0.0005% total nitrogen (N)

pH

5.5-7.5 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤50 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Foliar applied potassium nanoparticles (K-NPs) and potassium sulfate on growth, physiological, and phytochemical parameters in Melissa officinalis L. under salt stress.: This investigation assesses the effectiveness of potassium sulfate in alleviating salt stress in plants, focusing on its role in enhancing growth and stress tolerance. The study provides valuable data for improving crop resilience to salinity, a growing issue in global agriculture (Nanehkaran FM et al., 2024).

  • Effects of Potassium-Containing Fertilizers on Sugar and Organic Acid Metabolism in Grape Fruits.: Examining the role of potassium sulfate in grapevine nutrition, this research elucidates its impact on the metabolic pathways involved in sugar and organic acid production. The outcomes are significant for viticulture, offering strategies to enhance fruit quality and vine health (Wang J et al., 2024).

  • The combination of chemical fertilizer affected the control efficacy against root-knot nematode and environmental behavior of abamectin in soil.: This study investigates how potassium sulfate, when used in conjunction with other fertilizers, influences the efficacy of pesticides and the soil′s environmental dynamics. The findings are crucial for developing integrated pest management strategies that are environmentally sustainable (Jiang J et al., 2024).

Other Notes

Component of Pseudomonas Selective Agar Base, and Pseudomonas Agar P Base.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Xinjian Shi et al.
Nature communications, 5, 4775-4775 (2014-09-03)
Tungsten trioxide/bismuth vanadate heterojunction is one of the best pairs for solar water splitting, but its photocurrent densities are insufficient. Here we investigate the advantages of using helical nanostructures in photoelectrochemical solar water splitting. A helical tungsten trioxide array is
Souad Amiar et al.
Cell reports, 30(11), 3778-3792 (2020-03-19)
Apicomplexan parasites are unicellular eukaryotic pathogens that must obtain and combine lipids from both host cell scavenging and de novo synthesis to maintain parasite propagation and survival within their human host. Major questions on the role and regulation of each
Tian-Li Ma et al.
BMC plant biology, 12, 161-161 (2012-09-12)
Potassium (K+) is an important nutrient ion in plant cells and plays crucial roles in many plant physiological and developmental processes. In the natural environment, K+ deficiency is a common abiotic stress that inhibits plant growth and reduces crop productivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service