Skip to Content
Merck
All Photos(1)

Key Documents

60246

Sigma-Aldrich

Potassium formate

BioUltra, ≥99.0% (NT)

Synonym(s):

Formic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOK
CAS Number:
Molecular Weight:
84.12
Beilstein:
3623272
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (NT)

form

solid

impurities

insoluble matter, passes filter test

pH

7.0-8.5 (25 °C, 1 M in H2O)

mp

165-168 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.09
λ: 280 nm Amax: 0.08

SMILES string

[K+].[O-]C=O

InChI

1S/CH2O2.K/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

WFIZEGIEIOHZCP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium formate may be used with 1-propanol for the deproteinization and extraction of CoQ(10) and CoQ(10)H(2) from plasma. Potassium formate may be used as a plant photoinhibition prevention agent.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Shiraishi et al.
Journal of bioscience and bioengineering, 89(6), 564-568 (2005-10-20)
In plants, environmental adversity often leads to the formation of reactive oxygen species (ROS) because of excess light energy. We found that incubation of rice leaves under high light intensity together with CO2-depleted air seriously damaged photosynthesis; however, the application
Yukio Nohara et al.
Journal of fluorescence, 21(6), 2093-2100 (2011-06-08)
It was shown that ubiquinone (CoQ(10)) and ubiquinol (CoQ(10)H(2)) produce fluorescence products under alkaline conditions when reacted with 2-cyanoacetamide. The reaction mixture from CoQ(10) gave fluorescence with excitation and emission maximum wavelengths at 442 nm and 549 nm, respectively. This
Kyle C Costa et al.
mBio, 4(2), doi:10-doi:10 (2013-02-28)
Hydrogenotrophic methanogenic Archaea require reduced ferredoxin as an anaplerotic source of electrons for methanogenesis. H(2) oxidation by the hydrogenase Eha provides these electrons, consistent with an H(2) requirement for growth. Here we report the identification of alternative pathways of ferredoxin
Cláudia Freitas et al.
Applied biochemistry and biotechnology, 169(7), 2038-2048 (2013-01-30)
The use of lignocellulosic materials as substrate for bioethanol production is considered a cost-effective approach to make the biofuel production process economically sustainable. However, lignocellulosic hydrolysis releases toxic compounds such as weak acids which inhibit microorganism growth and ethanol production.
Kyle C Costa et al.
Journal of bacteriology, 195(7), 1456-1462 (2013-01-22)
Hydrogenotrophic methanogenic Archaea are defined by an H2 requirement for growth. Despite this requirement, many hydrogenotrophs are also capable of growth with formate as an electron donor for methanogenesis. While certain responses of these organisms to hydrogen availability have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service