Skip to Content
Merck
All Photos(1)

Key Documents

59980

Sigma-Aldrich

Jojoba oil from Simmondsia chinensis

tested according to DAC

Synonym(s):

Jojobae oelum, Jojoba bean oil, Jojoba liquid wax

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

density

0.87 g/mL at 20 °C

lipid type

oils

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Topical application of jojoba (Simmondsia chinensis L.) wax enhances the synthesis of pro-collagen III and hyaluronic acid and reduces inflammation in the ex-vivo human skin organ culture model.: This research demonstrates that jojoba wax can significantly enhance skin health by boosting collagen synthesis and reducing inflammation, making it a valuable ingredient in dermatological applications (Tietel et al., 2024).

  • Passive Enhancement of Retinol Skin Penetration by Jojoba Oil Measured Using the Skin Parallel Artificial Membrane Permeation Assay (Skin-PAMPA): A Pilot Study.: This pilot study shows that jojoba oil can enhance the penetration of retinol into the skin, suggesting its potential to improve the efficacy of topical retinol treatments (Gruber et al., 2023).

  • Anti-Herpes Simplex 1 Activity of Simmondsia chinensis (Jojoba) Wax.: This article reports on the antiviral properties of jojoba wax, specifically against Herpes Simplex Virus 1, providing a basis for its use in antiviral formulations (Tietel et al., 2021).

  • Inhalable Jojoba Oil Dry Nanoemulsion Powders for the Treatment of Lipopolysaccharide- or H(2)O(2)-Induced Acute Lung Injury.: This study investigates the use of jojoba oil in inhalable nanoemulsion powders for treating acute lung injury, highlighting its therapeutic potential in respiratory conditions (Zhang et al., 2021).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A case of contact dermatitis from jojoba.
L Di Berardino et al.
Contact dermatitis, 55(1), 57-58 (2006-07-18)
Andrea C Keenan et al.
Journal of cosmetic science, 62(2), 149-160 (2011-06-04)
The ethnic hair care market is large and diverse, with many unmet needs, especially when the definition of ethnic varies as much as the hair does. By examining the variety of hair care raw materials now available, we designed hair
Larissa Meier et al.
Forschende Komplementarmedizin (2006), 19(2), 75-79 (2012-05-16)
External application of clay facial masks is a cosmetic procedure generally used to reduce skin lesions and to improve overall skin condition. Collecting pilot data about self-treatment with clay jojoba oil masks on participants with acne-prone, lesioned skin and acne.
Mostafa Shahin et al.
AAPS PharmSciTech, 12(1), 239-247 (2011-01-13)
Jojoba oil-based emulgel formulations were prepared using different concentrations of various gelling agents, such as hydroxypropyl methylcellulose (HPMC) and Carbopol 934 P and combination of both. The prepared emulgels were physically evaluated for their stability after temperature cycle test, centrifugation
Manuel Gamez-Garcia
Journal of cosmetic science, 60(2), 85-95 (2009-05-20)
An analysis of the light interference patterns produced by the penetration and removal of lipids from the cuticle sheath has shown that cuticle cells in their virgin state have a few intrinsic imperfections in the form of voids and cracks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service