Skip to Content
Merck

58879

L-Isoleucine

≥99.5% (TLC), BioUltra

Synonym(s):

(2S,3S)-2-Amino-3-methylpentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-798-2
MDL number:
Beilstein/REAXYS Number:
1721792
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Isoleucine, BioUltra, ≥99.5% (NT)

product line

BioUltra

Quality Segment

assay

≥99.5% (NT), ≥99.5% (TLC)

form

powder or crystals

optical activity

[α]20/D +41.0±1°, c = 5% in 6 M HCl

impurities

insoluble matter, passes filter test, ≤0.5% foreign amino acids

ign. residue

0.05%

loss

0.05% loss on drying, 20 °C (HV)

color

white

mp

288 °C (dec.) (lit.)

solubility

1 M HCl: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg, sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, NH4+: ≤100 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.07, λ: 280 nm Amax: 0.05

SMILES string

CC[C@H](C)[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

Application


  • Enhanced protein-metabolite correlation analysis: To investigate the association between Staphylococcus aureus mastitis and metabolic immune pathways.: This article discusses the metabolic pathways associated with bacterial infections in livestock, potentially useful for chemists studying metabolic pathways and their linkage to immune responses (Chen et al., 2024).

  • Probing pharmaceutically important amino acids L-isoleucine and L-tyrosine Solubilities: Unraveling the solvation thermodynamics in diverse mixed solvent systems.: This study explores the solubility and solvation thermodynamics of l-isoleucine, providing critical data for chemists working with pharmaceutical formulations and solubility studies (Chakraborty et al., 2024).


Other Notes

Growth requirement of various microorganisms


Still not finding the right product?

Explore all of our products under L-Isoleucine


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library