Skip to Content
Merck

56449C

Minimum Essential Medium Joklik′s Modified

with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without calcium chloride, without sodium bicarbonate, dry powder, suitable for cell culture

Synonym(s):

Minimum Essential Medium Eagle, MEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

for research or for further manufacturing use

Quality Segment

form

dry powder

technique(s)

cell culture | mammalian: suitable

components

Earle’s salts (5% CO2): yes
NaHCO3: no
L-glutamine: 292 mg/L
phenol red: 10.620 mg/L

shipped in

ambient

storage temp.

2-8°C

Preparation Note

Formulated to contain 11.0 grams of powder per liter of medium.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library