Skip to Content
Merck
All Photos(1)

Key Documents

55540

Sigma-Aldrich

4-(Hydroxymercuri)benzoic acid sodium salt

≥95.0% (Hg)

Synonym(s):

p-Chloromercuribenzoate, p-Hydroxymercuribenzoic acid sodium salt, PCMB, Sodium 4-(hydroxymercurio)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5HgNaO3
CAS Number:
Molecular Weight:
360.69
Beilstein:
6118796
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95.0% (Hg)

mp

≥300 °C

SMILES string

[Na+].O[Hg]c1ccc(cc1)C([O-])=O

InChI

1S/C7H5O2.Hg.Na.H2O/c8-7(9)6-4-2-1-3-5-6;;;/h2-5H,(H,8,9);;;1H2/q;2*+1;/p-2

InChI key

CBELQCYFTSHKPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

4-(Hydroxymercuri)benzoic acid is an organic mercurial compound that reacts via mercury-sulfur affinity with sulfhydryl groups in peptides, proteins and other molecules. PCMB may be used for the determination of sulfhydryl groups in proteins according to Boyer, by atomic absorption, and stopped-flow measurements, G. Castronuovo-Lepore et al.. It may be used for specific applications such as probing the active site of an enzyme such as cytochrome C oxidase. 4-(Hydroxymercuri)benzoic acid appeared mistakenly as p-chloromercuriobenzoate [PCMB] in earlier literature.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mark D P Willcox et al.
Optometry and vision science : official publication of the American Academy of Optometry, 87(11), 839-846 (2010-09-08)
The purpose of this study was to investigate the effect of lens care products on short-term subjective and physiological performance silicone hydrogel lenses. Ten subjects wore either lotrafilcon B or galyfilcon A silicone hydrogel contact lenses soaked in a lens
Jörg R Aschenbach et al.
American journal of physiology. Gastrointestinal and liver physiology, 296(5), G1098-G1107 (2009-03-07)
The present study investigated the significance of apical transport proteins for ruminal acetate absorption and their interaction with different anions. In anion competition experiments in the washed reticulorumen, chloride disappearance rate (initial concentration, 28 mM) was inhibited by the presence
Cristian Follmer et al.
European journal of biochemistry, 271(7), 1357-1363 (2004-03-20)
In this work we compared two plant ureases, jackbean urease (JBU) and embryo-specific soybean urease (SBU) and a bacterial (Bacillus pasteurii) urease, for kinetic parameters and other biological properties described recently for ureases that are independent of the ureolytic activity.
Determination of protein thiol groups by atomic absorption spectrometry.
M Suzuki et al.
Analytical biochemistry, 32(1), 106-117 (1969-10-15)
Yulan Rao et al.
Journal of agricultural and food chemistry, 58(3), 1462-1468 (2010-01-13)
A liquid chromatography method with mass spectrometric detection has been developed for the simultaneous determination of six thiols in the sulfur metabolic pathway, including cysteine (Cys), homocysteine (HCys), glutathione (GSH), cysteinyl-glycine (Cys-Gly), gamma-glutamyl-cysteine (Glu-Cys), and S-adenosyl-homocysteine (AdoHcy). Tris(2-carboxyethyl)phosphine (TCEP) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service