Skip to Content
Merck

55463

Micro particles based on polystyrene, dark blue

size: 10 μm

Synonym(s):

Polystyrene blue microbeads (10 μm), Latex-coated polystyrene beads (10 μm), Latex beads from PS, dark blue

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

form

suspension (aqueous)

crosslinking

2 % cross-linked

concentration

5% (solids)

particle size

10 μm

bp

30-80 °C

mp

240 °C

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

General description

Ideal for profiling the particle size distribution (PSD) of a particle system.
The particle size of 10 μm is characterized using Coulter multisizer III.

Application

Used for standardization and validation of particle size analyzers.
Also used:
  • to produce a monolayer assembly of colloidal particles for optical microscopy study
  • as a model compound for comparison in uptake inhibition assay using A. castellanii trophozoites

Features and Benefits

  • suitable for routine instrument calibration checking, testing and corrections
  • particle size traceable to Community Bureau of Reference (BCR) standards
  • available in 5 mL pack size as a neat sample



Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



All-Aqueous Thin-Film-Flow-Induced Cell-Based Monolayers
Chan YK, et al.
ACS Applied Materials & Interfaces, 11(25), 22869-22877 (2019)
Toxoplasma gondii: uptake and survival of oocysts in free-living amoebae
Winiecka-Krusnell J, et al.
Experimental Parasitology, 121(2), 124-131 (2009)
Yau Kei Chan et al.
ACS applied materials & interfaces, 11(25), 22869-22877 (2019-05-31)
Cells in vitro usually require a solid scaffold to attach and form two-dimensional monolayer structures. To obtain a substrate-free cell monolayer, long culture time and specific detaching procedures are required. In this study, a thin-film-flow-induced strategy is reported to overcome



Global Trade Item Number

SKUGTIN
CRM05286-100MG04065265038317
45475-250MG04061833947845
55463-5ML-F04061837790645
55463-10ML-F04061837790638