Skip to Content
Merck
All Photos(1)

Key Documents

53467

Sigma-Aldrich

2,3-Dioleyloxy-1-(dimethylamino)propane

≥98.0% (TLC)

Synonym(s):

1,2-Dioleyloxy-3-(dimethylamino)propane, N,N-Dimethyl-2,3-bis[(9Z)-9-octadecen-1-yloxy]-1-propanamine, DODMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H81NO2
CAS Number:
Molecular Weight:
620.09
Beilstein:
10139013
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥98.0% (TLC)

form

liquid

functional group

amine
ether

lipid type

cationic lipids

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCOCC(CN(C)C)OCCCCCCCC\C=C/CCCCCCCC

InChI

1S/C41H81NO2/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-43-40-41(39-42(3)4)44-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h19-22,41H,5-18,23-40H2,1-4H3/b21-19-,22-20-

InChI key

GLGLUQVVDHRLQK-WRBBJXAJSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lloyd B Jeffs et al.
Pharmaceutical research, 22(3), 362-372 (2005-04-20)
A fully scalable and extrusion-free method was developed to prepare rapidly and reproducibly stabilized plasmid lipid particles (SPLP) for nonviral, systemic gene therapy. Liposomes encapsulating plasmid DNA were formed instantaneously by mixing lipids dissolved in ethanol with an aqueous solution
Yujing Li et al.
Nanomedicine : nanotechnology, biology, and medicine, 13(2), 371-381 (2016-10-18)
Microfluidic systems can accelerate clinical translation of nanoparticles due to their ability to generate nanoparticles in a well-controlled and reproducible manner. In this study, a single-step process based on microfluidic focusing (MF) was employed to synthesize transferrin-conjugated lipid nanoparticles (Tf-LNPs)
Adam Judge et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 13(2), 328-337 (2005-11-09)
The systemic application of nucleic acid drugs requires delivery systems that overcome the poor pharmacokinetics, limited biodistribution, and inefficient uptake of nucleic acids. PEGylated liposomes show considerable promise because of their intrinsic ability to accumulate at disease sites and facilitate
Cationic lipid saturation influences intracellular delivery of encapsulated nucleic acids.
Heyes, J., et al.
J. Controlled Release, 107, 276-287 (2005)
Brett A Duguay et al.
Journal of leukocyte biology, 104(3), 587-596 (2018-04-19)
Mast cells are important immune cells that have significant roles in mediating allergy and asthma. Therefore, studying the molecular mechanisms regulating these and other processes in mast cells is important to elucidate. Methods such as lipofection, transduction, and electroporation are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service