Skip to Content
Merck
All Photos(1)

Key Documents

53370

Sigma-Aldrich

L-Histidine monohydrochloride monohydrate

≥99.0% (AT)

Synonym(s):

L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4168261
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99.0% (AT)

form

solid

optical activity

[α]20/D +9.5±0.5°, c = 5% in 5 M HCl

impurities

≤0.3% foreign amino acids

mp

254 °C (dec.) (lit.)

solubility

H2O: 1 g/10 mL, clear, colorless (hot)

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤500 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Histidine is a hydrophilic essential amino acid that has a basic sidechain containing an imidazole group.

Biochem/physiol Actions

L-Histidine is biosynthesized from ATP, 5-phosphoribosyl-1-pyrophosphate (PRPP), and glutamine. This essential amino acid can be degraded to glutamate by histidiase, urocanase and imidazolonepropionase; it is also the precursor of histamine by action of histidine decarboxylase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemistry, 3rd. ed. (New York, NY)
Stryer, L., W.H. Freeman
Biochemistry (3rd Edition), 19-20 (1988)
Textbook of Biochemistry with Clinical Correlations, 5th ed. (New York, NY)
Devlin, T. M., ed., Wiley-Liss
Biochemistry (5th Edition), 97-97 (2002)
Sean E Lehman et al.
Journal of pharmaceutical sciences, 110(5), 1948-1957 (2021-01-17)
Protein aggregation can affect the quality of protein-based therapeutics. Attempting to unravel factors influencing protein aggregation involves systematic studies. These studies often include sodium azide or similar preservatives in the aggregation buffer. This work shows effects of azide on aggregation
Karla Viridiana Castro-Cerritos et al.
Bio-protocol, 7(23), e2634-e2634 (2017-12-05)
Elucidating how a population of non-growing bacteria generates mutations improves our understanding of phenomena like antibiotic resistance, bacterial pathogenesis, genetic diversity and evolution. To evaluate mutations that occur in nutritionally stressed non-growing bacteria, we have employed the strain B. subtilis
Martin Maldonado et al.
Neurochemistry international, 85-86, 31-39 (2015-05-06)
The effect of reserpine on histamine (HA) and tele-methylhistamine (N(τ)-MHA) in hypothalamus and cortex of rats was analyzed and compared to catecholamines. IP injection of reserpine (5 mg/kg) confirmed the effectiveness of reserpine treatment on noradrenaline and dopamine levels. Our in-vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service