Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥300 U/g
Biological source:
fungus (Rhizomucor miehei)
biological source
fungus (Rhizomucor miehei)
Quality Segment
form
powder
specific activity
≥300 U/g
storage temp.
2-8°C
Application
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Biochem/physiol Actions
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
The lipase cleaves ester bonds of triacylglycerols with the subsequent release of free fatty acids, diacylglycerols, monoacylglycerols and glycerol. It can also catalyze the reverse reactions when the aqueous medium is replaced by an organic or a biphasic aqueous/organic medium.
Other Notes
1 U corresponds to the amount of enzyme which liberates 1 μmol butyric acid per minute at pH 7.5 and 40°C (tributyrin, Cat. No. 91010, as substrate)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.