Skip to Content
Merck
All Photos(1)

Key Documents

46940

Sigma-Aldrich

6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester

suitable for fluorescence, ≥75% (HPLC)

Synonym(s):

Fluorescein-5(6)-carboxamidocaproic acid N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H26N2O10
CAS Number:
Molecular Weight:
586.55
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Assay

≥75% (HPLC)

form

solid

solubility

DMF: soluble
methanol: soluble

fluorescence

λex 490 nm; λem 514 nm (pH 8.2)
λex 491 nm; λem 515 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)C(=O)NCCCCCC(=O)ON6C(=O)CCC6=O)c1.Oc7ccc8c(Oc9cc(O)ccc9C8%10OC(=O)c%11ccc(cc%10%11)C(=O)NCCCCCC(=O)ON%12C(=O)CCC%12=O)c7

InChI

1S/2C31H26N2O10/c34-18-6-9-22-24(15-18)41-25-16-19(35)7-10-23(25)31(22)21-8-5-17(14-20(21)30(40)42-31)29(39)32-13-3-1-2-4-28(38)43-33-26(36)11-12-27(33)37;34-18-6-9-21-24(15-18)41-25-16-19(35)7-10-22(25)31(21)23-14-17(5-8-20(23)30(40)42-31)29(39)32-13-3-1-2-4-28(38)43-33-26(36)11-12-27(33)37/h2*5-10,14-16,34-35H,1-4,11-13H2,(H,32,39)

InChI key

WAGYELYAQOYZFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-[Fluorescein-5(6)-carboxamido] hexanoic acid N-hydroxysuccinimide ester (FNHS) is a fluorescent label, also known as fluorescein. FNHS max. excitation is at 490 nm with emission max. at 514nm. 6-[Fluorescein-5(6)-carboxamido] hexanoic acid N-hydroxysuccinimide ester binds covalently with primary amines.

Application

6-[Fluorescein-5(6)-carboxamido] hexanoic acid N-hydroxysuccinimide ester (FHS) is used as a reagent to conjugate the fluorophore 6-[Fluorescein-5(6)-carboxamido] hexanoic acid to molecules.6-[Fluorescein-5(6)-carboxamido] hexanoic acid is used as a fluorescein-based fluorescent label for proteins that can be analyzed in applications such as quenching experiments.
Reagent for introducing the fluorescein label; the spacer renders the reactive group more accessible to nucleophiles on biopolymers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Vaz et al.
Colloids and surfaces. B, Biointerfaces, 114, 11-19 (2013-10-29)
In this work, we present a suitable methodology to produce magnetically recoverable bioreactors based on enzymes, which are covalently attached on the surface of iron oxide@silica nanoparticles. In order to produce this system, iron oxide clusters with a mean diameter
R Bertrand et al.
Biochemistry, 34(29), 9500-9507 (1995-07-25)
We describe, for the first time, the reaction of skeletal myosin subfragment 1 (S-1) with the succinimido ester of 6-[fluorescein-5(and 6)-carboxamido]hexanoic acid (FHS), which takes place at pH 7.0, 20 degrees C, within a 15 min period, in the presence
J J MacLean et al.
Biophysical journal, 78(3), 1441-1448 (2000-02-29)
Lys-553 of skeletal muscle myosin subfragment 1 (S1) was specifically labeled with the fluorescent probe FHS (6-[fluorescein-5(and 6)-carboxamido]hexanoic acid succinimidyl ester) and fluorescence quenching experiments were carried out to determine the accessibility of this probe at Lys-553 in both the
Abdul Qader Abbady et al.
Frontiers in physiology, 8, 317-317 (2017-06-06)
Programed cell death is a critical and unavoidable part of life. One of the most widely used markers for dying cells, by apoptosis or pyroptosis, is the redistribution of phosphatidylserine (PS) from the inner to the outer plasma membrane leaflet.
Avidin binding of carboxyl-substituted biotin and analogues.
K Hofmann et al.
Biochemistry, 21(5), 978-984 (1982-03-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service