Skip to Content
Merck
All Photos(1)

Key Documents

41986

Sigma-Aldrich

Atto 550 azide

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352125
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

>90% (HPLC)

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm
550 nm

fluorescence

λex 554 nm; λem 576 nm±10 nm in 0.1 M phosphate pH 7.0

λ

(ethanol with 0.1% trifluoroacetic acid)

UV absorption

λ: 553-559 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smart-aggregation imaging for single molecule localization with SPAD cameras.
Gyongy, I.; et al.
arXiv (2016)
Rula Masoud et al.
ACS nano, 6(7), 6272-6283 (2012-06-06)
We present a test case example of a detailed single-molecule fluorescence study of one of the most sophisticated and complex DNA devices introduced to date, a recently published autonomous bipedal DNA motor. We used the diffusion-based single-molecule Förster resonance energy
Rahul Roy et al.
Nature methods, 5(6), 507-516 (2008-05-31)
Single-molecule fluorescence resonance energy transfer (smFRET) is one of the most general and adaptable single-molecule techniques. Despite the explosive growth in the application of smFRET to answer biological questions in the last decade, the technique has been practiced mostly by
Robert H Meltzer et al.
Lab on a chip, 11(5), 863-873 (2011-01-21)
Rapid, specific, and sensitive detection of airborne bacteria, viruses, and toxins is critical for biodefense, yet the diverse nature of the threats poses a challenge for integrated surveillance, as each class of pathogens typically requires different detection strategies. Here, we
A molecular toolkit for population genetic investigations of the ash dieback pathogen Hymenoscyphus pseudoalbidus.
Gross, A., et al.
Forest Pathology, 42, 252-264 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service