Skip to Content
Merck

41573

Trizma® hydrochloride buffer solution

BioUltra, Molecular Biology, pH 7.2

Synonym(s):

Tris(hydroxymethyl)aminomethane hydrochloride buffer solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3675235
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

product line

BioUltra

form

liquid

impurities

DNases, RNases, none detected, insoluble matter, passes filter test, proteases, phosphatases, none detected

pH

7.2±0.1 (25)

density

1.05 g/mL at 20 °C

cation traces

Al: ≤1 mg/kg, As: ≤0.1 mg/kg, Ba: ≤1 mg/kg, Bi: ≤1 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤20 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Na: ≤20 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03, λ: 280 nm Amax: 0.04

application(s)

diagnostic assay manufacturing

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
41573-1L-F04061838169181