Skip to Content
Merck
All Photos(1)

Key Documents

41111

Sigma-Aldrich

Ajmalicine

≥98.0% (HPLC)

Synonym(s):

δ-Yohimbine, Py-Tetrahydroserpentine, Raubasine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N2O3
CAS Number:
Molecular Weight:
352.43
Beilstein:
97268
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D -65±3°, c = 1 in chloroform

mp

~258 °C (dec.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

N21[C@@H](C[C@H]5[C@@H]([C@@H](OC=C5C(=O)OC)C)C2)c3[nH]c4c(c3CC1)cccc4

InChI

1S/C21H24N2O3/c1-12-16-10-23-8-7-14-13-5-3-4-6-18(13)22-20(14)19(23)9-15(16)17(11-26-12)21(24)25-2/h3-6,11-12,15-16,19,22H,7-10H2,1-2H3/t12-,15-,16+,19-/m0/s1

InChI key

GRTOGORTSDXSFK-XJTZBENFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ajmalicine (δ-Yohimbine, Py-Tetrahydroserpentine, Raubasine) is an alkaloid used to study its effects as an antagonist of adrenergic and nicotinic receptors.

Biochem/physiol Actions

Metabolite in the indole alkaloid biosynthesis (serpentine production); found naturally in various plants such as Rauwolfia spp., Catharanthus roseus, and Mitragyna speciosa. It shows antimicrobial activity, and is used as an anti-hypertensive and sedative.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W G Kurz et al.
Planta medica, 42(1), 22-31 (1981-05-01)
A cell line of Catharanthus roseus (L.) G. Don coded PRL # 200, was characterized with respect to its biosynthetic capabilities for indolealkaloids, in particular catharanthine, in suspension cultures. Other alkaloids isolated are vallesiachotamine isomers, ajmalicine, hörhammericine, hörhammerinine, vindolinine, 19-epivindolinine
Roberts, M. F.
Alkaloids: biochemistry, ecology, and medicinal applications, 450-450 (1998)
David M Pereira et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 17(8-9), 646-652 (2009-12-08)
The leaves of Catharanthus roseus constitute the only source of the well known indolomonoterpenic alkaloids vincristine and vinblastine. In this work we studied the biological potential of the roots, which are used in several countries as decocts or hot water
Francisco León et al.
Natural product communications, 4(7), 907-910 (2009-09-08)
Mitragyna speciosa (Rubiaceae) has traditionally been used in the tropical regions of Asia, Africa and Indonesia as a substitute for opium. Indole alkaloids are the most common compounds that have been isolated. We investigated the constituents of the leaves of
Catharanthus alkaloids and their enhanced production using elicitors: a review.
Gautam, S., et al.
International Journal of Pharmacy and Technology, 3, 713-724 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service