Skip to Content
Merck

3951

Periodic Acid Solution

1 g/dL in deionized water

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Level

shelf life

expiry date on label

reaction suitability

reagent type: oxidant

IVD

for in vitro diagnostic use

concentration

1 g/dL in deionized water

pH

1.77

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OI(O)(O)(O)(O)=O

InChI

1S/H5IO6/c2-1(3,4,5,6)7/h(H5,2,3,4,5,6,7)

InChI key

TWLXDPFBEPBAQB-UHFFFAOYSA-N

Application

For use in procedure no. 395 for Periodic Acid-Schiff (PAS) and Silver Stain Procedure No. HT100.


Still not finding the right product?

Explore all of our products under Periodic Acid Solution


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 2

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Allison M Bradbury et al.
Journal of neuroscience research, 94(11), 1007-1017 (2016-09-18)
Globoid cell leukodystrophy (GLD), or Krabbe's disease, is a debilitating and always fatal pediatric neurodegenerative disease caused by a mutation in the gene encoding the hydrolytic enzyme galactosylceramidase (GALC). In the absence of GALC, progressive loss of myelin and accumulation
Robert W Morton et al.
The Journal of physiology, 597(17), 4601-4613 (2019-07-12)
Performing resistance exercise with heavier loads is often proposed to be necessary for the recruitment of larger motor units and activation of type II muscle fibres, leading to type II fibre hypertrophy. Indirect measures [surface electromyography (EMG)] have been used
Sonja Frimmel et al.
Journal of ophthalmic & vision research, 12(2), 175-182 (2017-05-26)
Diabetic retinopathy is a leading cause of vision loss. There is a great need for early diagnosis prior to the occurrence of irreversible structural damages. Expression of endothelial adhesion molecules is observed before the onset of diabetic vascular damage; however



Global Trade Item Number

SKUGTIN
27130-10G04061826143759
3951-100ML04061831982961