Skip to Content
Merck

Skip To

38790

(±)-Pinostrobin

≥99.0% (TLC)

Synonym(s):

(RS)-2,3-Dihydro-5-hydroxy-7-methoxy-2-phenyl-4H-1-benzopyran-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Assay:
≥99.0% (TLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (TLC)

SMILES string

COc1cc(O)c2C(=O)CC(Oc2c1)c3ccccc3

InChI

1S/C16H14O4/c1-19-11-7-12(17)16-13(18)9-14(20-15(16)8-11)10-5-3-2-4-6-10/h2-8,14,17H,9H2,1H3

InChI key

ORJDDOBAOGKRJV-UHFFFAOYSA-N

Application

(±)-Pinostrobin, a chiral flavonoid (±), may be used in the development of analytical methods for separation of its individual enantiomers. Pinostrobin may be used to study its neuroprotective antioxidant effects.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
41934PHL80061PHL84060
assay

≥99.0% (TLC)

assay

≥98% (HPLC)

assay

≥98.0% (HPLC)

assay

≥90.0% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters